Skip to Content
Merck
All Photos(1)

Key Documents

O806

Sigma-Aldrich

1-Octadecene

technical grade, 90%

Synonym(s):

ODE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH=CH2
CAS Number:
Molecular Weight:
252.48
Beilstein:
1761067
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

0.71 (vs air)

Assay

90%

autoignition temp.

482 °F

refractive index

n20/D 1.444 (lit.)

bp

314.4 +/- 5.0 °C/760 mmHg (lit.)
179 °C/15 mmHg (lit.)
314.4 °C±5.0 °C/760 mmHg (lit.)

mp

14-16 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCC=C

InChI

1S/C18H36/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3H,1,4-18H2,2H3

InChI key

CCCMONHAUSKTEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octadecene is a long-chain hydrocarbon used in the synthesis of nanoparticles as a capping and reducing agent.

Application

1-Octadecene has been commonly used as a non-coordinating solvent for growing nanoparticles and quantum dots due to its high boiling point. The 1-octadecene monolayer is a promising passivating coating in nanoelectronics due to its stability and chemical protection against oxidation, effects of acids and bases without altering the properties.

Other Notes

Contains 2-octyl-1-decene, n-octadecane, 2-butyl-1-tetradecene, 2-hexyl-1-dodecene

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrical passivation of the silicon surface by organic monolayers of 1-octadecene.
Antonova IV
Semiconductors, 41(8), 991-997 (2007)
Xiao-Shuai Zhang et al.
Journal of nanobiotechnology, 16(1), 42-42 (2018-04-21)
Ag2S has the characteristics of conventional quantum dot such as broad excitation spectrum, narrow emission spectrum, long fluorescence lifetime, strong anti-bleaching ability, and other optical properties. Moreover, since its fluorescence emission is located in the NIR-II region, has stronger penetrating
Photoluminescent colloidal CdS nanocrystals with high quality via noninjection one-pot synthesis in 1-octadecene.
Ouyang J
The Journal of Physical Chemistry C, 113(18), 7579-7593 (2009)
Wei Zheng et al.
Nature communications, 9(1), 3462-3462 (2018-08-29)
All-inorganic CsPbX3 (X = Cl, Br, and I) perovskite quantum dots (PeQDs) have shown great promise in optoelectronics and photovoltaics owing to their outstanding linear optical properties; however, nonlinear upconversion is limited by the small cross-section of multiphoton absorption, necessitating high power
The effectiveness of electroporation-based nanocurcumin and curcumin treatments on human breast cancer cells.
Lin W
Proceedings of ESA annual meeting on electrostatics, 17-19 (2014)

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service