Skip to Content
Merck
All Photos(2)

Documents

O8880

Sigma-Aldrich

Oleyl alcohol

≥99% (GC)

Synonym(s):

Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2OH
CAS Number:
Molecular Weight:
268.48
Beilstein:
1723962
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (GC)

refractive index

n20/D 1.46 (lit.)

bp

207 °C/13 mmHg (lit.)

mp

0-5.0 °C (lit.)

density

0.849 g/mL at 25 °C (lit.)

lipid type

unsaturated FAs

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCO

InChI

1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3/b10-9-

InChI key

ALSTYHKOOCGGFT-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eunsung Kan et al.
Biotechnology and bioengineering, 102(3), 708-713 (2008-10-25)
The sensitivity of a conceptual model of a foam emulsion bioreactor (FEBR) used for the control of toluene vapors in air was examined. Model parametric sensitivity studies showed which parameters affect the removal of toluene (as model pollutant) in the
Shiho Yamashita et al.
Applied microbiology and biotechnology, 74(4), 761-767 (2006-11-24)
Rhodococcus opacus strain B-4, which has recently been isolated as an organic solvent-tolerant bacterium, has a high hydrophobicity and exhibits a high affinity for hydrocarbons. This bacterium was able to survive for at least 5 days in organic solvents, including
Rupali Dhara et al.
Journal of oleo science, 59(4), 169-176 (2010-03-20)
The amount and characterization of phytosterol and other minor components present in three Indian minor seed oils, mahua (Madhuca latifolia), sal (Shorea robusta) and mango kernel (Mangifera indica), have been done. Theses oils have shown commercial importance as cocoa-butter substitutes
Emily A Simpson et al.
Physical chemistry chemical physics : PCCP, 12(37), 11565-11575 (2010-08-18)
This paper presents the studies of one and two component particles using a CO(2) laser for vaporization and VUV ionization in an ion trap mass spectrometer. The degree of fragmentation for a one component system was demonstrated to be a
Klaus Ejner Andersen et al.
Contact dermatitis, 55(6), 354-356 (2006-11-15)
We report an atopic dermatitis patient with recurrent hand dermatitis who developed a severe allergic contact dermatitis from the use of Elidel cream. Diagnostic patch tests showed an isolated contact allergy to the emulsifier oleyl alcohol present in the product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service