Skip to Content
Merck
All Photos(1)

Key Documents

33610

Sigma-Aldrich

(+)-2,3-Dibenzoyl-D-tartaric acid

≥99.0% (T)

Synonym(s):

(+)-O,O′-Dibenzoyl-D-tartaric acid, D-Tartaric acid 2,3-dibenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5CO2CH(CO2H)]2
CAS Number:
Molecular Weight:
358.30
Beilstein:
2227343
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (T)

form

powder

optical activity

[α]20/D +117±2°, c = 5% in ethanol

impurities

≤1% water

mp

145-155 °C
154-156 °C (lit.)

functional group

carboxylic acid
ester
phenyl

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](OC(=O)c1ccccc1)[C@H](OC(=O)c2ccccc2)C(O)=O

InChI

1S/C18H14O8/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12/h1-10,13-14H,(H,19,20)(H,21,22)/t13-,14-/m0/s1

InChI key

YONLFQNRGZXBBF-KBPBESRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-2,3-Dibenzoyl-D-tartaric acid is an acidic chiral resolving reagent for the resolution of bases.

Other Notes

Reagent for the resolution of bases

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical continuous resolution of a-amino-e-caprolactam by diastereomeric salt formation using a single resolving agent with a solvent switch method.
Sakai K, et al.
Tetrahedron Asymmetry, 14(23), 3713-3718 (2003)
Mirjan M van Timmeren et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 20(11), 2349-2357 (2005-09-08)
Non-esterified fatty acids (NEFA) carried on albumin may have a causal role in the development of chronic proteinuria-induced nephropathy. To investigate whether NEFA aggravate renal structural damage, we studied the effects of NEFA addition to delipidated bovine serum albumin (BSA)
S. Collina et al.
Tetrahedron Asymmetry, 10, 2387-2387 (1999)
M.R. Caira et al.
Tetrahedron Asymmetry, 10, 2175-2175 (1999)
P.H. Boyle
Quart. Rev., 25, 323-323 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service