Skip to Content
Merck
All Photos(1)

Key Documents

C2107

Sigma-Aldrich

(1S)-(+)-10-Camphorsulfonic acid

99%

Synonym(s):

(+)-Camphor-10-sulfonic acid (β), (1S)-Camphor-10-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Beilstein:
2809675
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

crystals

optical activity

[α]20/D +19.9°, c = 2 in H2O

mp

196-200 °C (dec.) (lit.)

SMILES string

CC1(C)[C@@H]2CC[C@@]1(CS(O)(=O)=O)C(=O)C2

InChI

1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m1/s1

InChI key

MIOPJNTWMNEORI-GMSGAONNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(1S)-(+)-10-Camphorsulfonic acid may be used as a starting material to synthesize 10-iodocamphor.[1] It may also be used as a dopant to induce chirality in the conduction band of polyaniline.[2] (±)-S-methyl-S-phenylsulfoximine can be resolved using it as a chiral resolving agent to form the (-)-form in high enantiomeric purity.[3]
Used as a resolving agent,[4] as a catalyst for coupling dipeptides.[5]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Large induced optical activity in the conduction band of polyaniline doped with (1S)-(+)-10-camphorsulfonic acid.
Havinga EE, et al.
Synt. Metals, 66(1), 93-97 (1994)
Norma J Greenfield
Nature protocols, 1(6), 2876-2890 (2007-04-05)
Circular dichroism (CD) is an excellent tool for rapid determination of the secondary structure and folding properties of proteins that have been obtained using recombinant techniques or purified from tissues. The most widely used applications of protein CD are to
Heterocycles, 31, 353-353 (1990)
Helvetica Chimica Acta, 69, 1153-1153 (1986)
An efficient resolution of (?)-S-methyl-S-phenylsulfoximine with (+)-10-camphorsulfonic acid by the method of half-quantities.
Brandt J and Gais HJ.
Tetrahedron Asymmetry, 8(6), 909-912 (1997)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service