Skip to Content
Merck
All Photos(1)

Documents

163449

Sigma-Aldrich

(+)-2,3-Dibenzoyl-D-tartaric acid

≥98%, made from synthetic tartaric acid

Synonym(s):

(+)-O,O′-Dibenzoyl-D-tartaric acid, D-Tartaric acid 2,3-dibenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5CO2CH(CO2H)]2
CAS Number:
Molecular Weight:
358.30
Beilstein:
2227343
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

optical activity

[α]28/D +116°, c = 9 in ethanol

mp

154-156 °C (lit.)

SMILES string

OC(=O)[C@@H](OC(=O)c1ccccc1)[C@H](OC(=O)c2ccccc2)C(O)=O

InChI

1S/C18H14O8/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12/h1-10,13-14H,(H,19,20)(H,21,22)/t13-,14-/m0/s1

InChI key

YONLFQNRGZXBBF-KBPBESRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-2,3-Dibenzoyl-D-tartaric acid is commonly used as an optically active resolving agent in the chiral resolution process such as diastereomeric salt resolution technique.

Application

Reagent for chiral resolution of amino compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 2, 989-989 (1991)
Analysis and Comparison of Commonly Used Acidic Resolving Agents in Diastereomeric Salt Resolution-Examples for dl-Serine.
Sistla VS, et al.
Crystal Growth & Design, 11(9), 3761-3768 (2011)
Emily M Jutkiewicz et al.
The Journal of pharmacology and experimental therapeutics, 309(1), 173-181 (2004-01-15)
The diarylpiperazine delta-opioid agonist SNC80 [(+)-4-[(alphaR)-alpha-[(2S,5R)-2,5-dimethyl-4-(2-propenyl)-1-piperazinyl]-(3-methoxyphenyl)methyl]-N,N-diethylbenzamide] produces convulsions, antidepressant-like effects, and locomotor stimulation in rats. The present study compared the behavioral effects in Sprague-Dawley rats of SNC80 with its two derivatives, SNC86 [(+)-4-[alpha(R)-alpha-[(2S,5R)-2,5-dimethyl-4-(2-propenyl)-1-piperazinyl]-(3-hydroxyphenyl)methyl]-N,N-diethylbenzamide] and SNC162 [(+)-4-[(alphaR)-alpha-[(2S,5R)-2,5-dimethyl-4-(2-propenyl)-1-piperazinyl]-(3-phenyl)methyl]-N,N-diethylbenzamide], which differ by one
C Q Cao et al.
European journal of pharmacology, 418(1-2), 79-87 (2001-05-04)
Due to low central nervous system (CNS) bioavailability of delta-opioid peptides, little is known about the effect of systemic administration of delta-opioid receptor ligands. The present study examined the effect of non-peptidergic delta-opioid receptor agonists, (+)-4-[(alphaR)-alpha-((2R,5R)-4-Allyl-2,5-dimethyl-1-piperazinyl)-3-methoxybenzyl]-N,N-diethylbenzamide (SNC80) and (-)dibenzoyl-L-tartaric acid
Jianghong An et al.
Journal of medicinal chemistry, 52(9), 2667-2672 (2009-05-08)
Computational molecular docking provides an efficient and innovative approach to examine small molecule and protein interactions. We have utilized this method to identify potential inhibitors of the H5N1 neuraminidase protein. Of the 20 compounds tested, 4-(4-((3-(2-amino-4-hydroxy-6-methyl-5-pyrimidinyl)propyl)amino)phenyl)-1-chloro-3-buten-2-one (1) (NSC89853) demonstrated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service