Skip to Content
Merck
All Photos(1)

Key Documents

282146

Sigma-Aldrich

(1R)-(−)-10-Camphorsulfonic acid

98%

Synonym(s):

(−)-Camphor-10-sulfonic acid, (1R)-(−)-Camphor-10-sulfonic acid, (1R)-Camphor-10-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Beilstein:
2809676
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

optical activity

[α]20/D −21°, c = 2 in H2O

mp

198 °C (dec.) (lit.)

functional group

ketone
sulfonic acid

SMILES string

[H][C@]12CC[C@](CS(O)(=O)=O)(C(=O)C1)C2(C)C

InChI

1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m0/s1

InChI key

MIOPJNTWMNEORI-XVKPBYJWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1R)-(−)-10-Camphorsulfonic acid is an HPLC derivatization reagent for UV/Vis detection. It is mainly employed for the resolution of bases.

Application

(1R)-(−)-10-Camphorsulfonic acid may be used as a chiral building block for the synthesis of pentavalent organo-bismuth derivatives by enantioselective C-arylation.[1] It may be used as a chiral monomer in the enantioselective sensing of chiral amino acids by potentiometric sensors based on optical active polyaniline films.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective sensing of chiral amino acids by potentiometric sensors based on optical active polyaniline films
Yin X, et al.
Biosensors And Bioelectronics, 21(11), 2184-2187 (2006)
Synthesis and reactivity of chiral pentavalent bismuth derivatives
Finet PJ and Fedorov YA
Russian Chemical Bulletin, 53(7), 1488-1495 (2004)
Katarzyna Jankowska et al.
Environmental research, 184, 109332-109332 (2020-03-11)
Novel electrospun poly(methyl methacrylate)/polyaniline electrospun fibres were produced, characterised, modified, and used as a support for laccase immobilisation by two methods: adsorption and covalent binding. Effective deposition of laccase by both methods was confirmed by FTIR and CLSM results. Nevertheless
Anne-Catherine Servais et al.
Electrophoresis, 25(16), 2701-2710 (2004-09-08)
The enantiomeric separation of a series of basic pharmaceuticals (beta-blockers, local anesthetics, sympathomimetics) has been investigated in nonaqueous capillary electrophoresis (NACE) systems using heptakis(2,3-di-O-methyl-6-O-sulfo)-beta-cyclodextrin (HDMS-beta-CD) in combination with potassium camphorsulfonate (camphorSO3-). For this purpose, a face-centered central composite design with
Kaoru Nobuoka et al.
Physical chemistry chemical physics : PCCP, 9(44), 5891-5896 (2007-11-09)
We directly observe the interaction between 1-butyl-3-methylimidazolium (bmim) or 1-butyl-2,3-dimethylimidazolium (bm(2)im) and the solute, ethyl acrylate (EA), which is the popular dienophile in the Diels-Alder reaction and an H-bonding acceptor, by using specially designed electrospray mass spectrometry. In imidazolium ionic

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service