Skip to Content
Merck
All Photos(3)

Key Documents

301124

Sigma-Aldrich

Lithium dicyclohexylamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2NLi
CAS Number:
Molecular Weight:
187.25
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

SMILES string

[Li]N(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22N.Li/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;/h11-12H,1-10H2;/q-1;+1

InChI key

HTZGVHYSMVGNOV-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Sol. 1 - Skin Corr. 1B - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service