Skip to Content
Merck
All Photos(1)

Key Documents

774766

Sigma-Aldrich

Lithium diisopropylamide solution

1.0 M in THF/hexanes

Synonym(s):

LDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NLi
CAS Number:
Molecular Weight:
107.12
Beilstein:
3655042
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in THF/hexanes

density

0.719 g/mL at 25 °C

functional group

amine

storage temp.

2-8°C

SMILES string

[Li]N(C(C)C)C(C)C

InChI

1S/C6H14N.Li/c1-5(2)7-6(3)4;/h5-6H,1-4H3;/q-1;+1

InChI key

ZCSHNCUQKCANBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This material is in a solution of THF/Hexanes (ca. 1:7 ratio, respectively)

Application

Titanium-ate enolates prepared from ketones by Ti(OiPr)4 and LDA underwent efficient radical trifluoromethylation providing trifluoromethyl ketones in good yield. Used in directed ortho-lithiation of arylsulfonyloxazolidinones to prepare N-substituted saccharin analogs.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 6405-6405 (2006)
Joshua D Schaefferkoetter et al.
Physics in medicine and biology, 60(14), 5543-5556 (2015-07-03)
In the context of investigating the potential of low-dose PET imaging for screening applications, we developed methods to assess small lesion detectability as a function of the number of counts in the scan. We present here our methods and preliminary
Juan Eugenio Iglesias et al.
NeuroImage, 115, 117-137 (2015-05-06)
Automated analysis of MRI data of the subregions of the hippocampus requires computational atlases built at a higher resolution than those that are typically used in current neuroimaging studies. Here we describe the construction of a statistical atlas of the
Martin E Gosnell et al.
Biochimica et biophysica acta, 1863(1), 56-63 (2015-10-04)
Hyperspectral imaging uses spectral and spatial image information for target detection and classification. In this work hyperspectral autofluorescence imaging was applied to patient olfactory neurosphere-derived cells, a cell model of a human metabolic disease MELAS (mitochondrial myopathy, encephalomyopathy, lactic acidosis
Mogens K Boisen et al.
PloS one, 9(10), e109430-e109430 (2014-10-21)
We tested the hypothesis that expression of microRNAs (miRNAs) in cancer tissue can predict effectiveness of bevacizumab added to capecitabine and oxaliplatin (CAPEOX) in patients with metastatic colorectal cancer (mCRC). Patients with mCRC treated with first line CAPEOX and bevacizumab

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service