Skip to Content
Merck
All Photos(2)

Key Documents

578940

Sigma-Aldrich

Tri-tert-butylphosphonium tetrafluoroborate

97%

Synonym(s):

TTBP · HBF4, [(t-Bu)3PH]BF4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3C]3PH]BF4
CAS Number:
Molecular Weight:
290.13
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Carbonylations

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

261 °C (lit.)

functional group

phosphine

SMILES string

F[B-](F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P.BF4/c1-10(2,3)13(11(4,5)6)12(7,8)9;2-1(3,4)5/h1-9H3;/q;-1/p+1

InChI key

YTJUCJAUJCXFTN-UHFFFAOYSA-O

Application

Hindered Phosphine salt employed with a Pd(0)-15-membered, triolefinic, macrocyle in Suzuki cross-coupling reactions of aryl bromides and chlorides. Also used in Heck coupling of non-activated vinyl tosylates with electron deficient olefins.
Ligand used in the Pd-catalyzed enantioselective α−arylation of N-boc-pyrrolidine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heck coupling with nonactivated alkenyl tosylates and phosphates: examples of effective 1,2-migrations of the alkenyl palladium(II) intermediates.
Anders L Hansen et al.
Angewandte Chemie (International ed. in English), 45(20), 3349-3353 (2006-04-12)
Kevin R Campos et al.
Journal of the American Chemical Society, 128(11), 3538-3539 (2006-03-16)
This communication discloses the first instance of the enantioselective Pd-catalyzed alpha-arylation of N-Boc-pyrrolidine. The methodology relies on Beak's sparteine-mediated, enantioselective deprotonation of N-Boc-pyrrolidine to form the 2-pyrrolidinolithium specices in high enantiomeric ratio (er). Transmetalation of this intermediate with zinc chloride
Synlett, 3001-3001 (2006)

Articles

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service