Skip to Content
Merck
All Photos(4)

Key Documents

202614

Sigma-Aldrich

Aluminum sulfate

99.99% trace metals basis

Synonym(s):

Aluminium sulphate, Aluminum alum, Dialuminum sulfate, dialuminum trisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3
CAS Number:
Molecular Weight:
342.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

impurities

≤150.0 ppm Trace Metal Analysis

mp

770 °C (dec.) (lit.)

density

2.71 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

DIZPMCHEQGEION-UHFFFAOYSA-H

General description

Aluminum sulfate is a white crystalline solid that is highly soluble in water. It is thermally stable and has the ability to act as a coagulant and flocculant. It is widely used in the field of solar cells, water treatment and batteries.

Application

Aluminum sulfate can be used:
  • As a surface modification agent/co-adsorbent for dye sensitizers in solar cell. It improves the surface of photoelectrode and purify dye solution through flocculation.
  • As an electrolyte additive for lead-acid batteries to effectively inhibit the passivation of electrodes. It helps to slow down cathode sulfation and improve the reversibility of the electrode reaction.
  • To prepare aqueous electrolyte for high energy aluminum electrochemical cells. It helps to form artificial solid electrolyte interphase to attain high specific energy.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Basic aluminum sulfate@ graphene hydrogel composites: preparation and application for removal of fluoride.
Chen Y, et al.
Journal of Material Chemistry A, 1(42), 13101-13110 (2013)
Clint S Schmidt et al.
Vaccine, 30(52), 7594-7600 (2012-10-27)
The investigational vaccine, NDV-3, contains the N-terminal portion of the Candida albicans agglutinin-like sequence 3 protein (Als3p) formulated with an aluminum hydroxide adjuvant in phosphate-buffered saline. Preclinical studies demonstrated that the Als3p vaccine antigen protects mice from oropharyngeal, vaginal and
Decarbonylation of lactic acid to acetaldehyde over aluminum sulfate catalyst.
Zhai Z, et al.
Industrial & Engineering Chemistry Research, 53(25), 10318-10327 (2014)
A statistical experiment design approach for arsenic removal by coagulation process using aluminum sulfate.
Baskan MB and Pala A
Desalination, 254(1), 42-48 (2010)
An easy, reproducible and cost-effective method for andrologists to improve the laboratory diagnosis of non-obstructive azoospermia: a novel microcentrifugation technique.
International Brazilian Journal of Urology : Official Journal of the Brazilian Society of Urology, 42(1), 132-138 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service