Skip to Content
Merck
All Photos(3)

Key Documents

368458

Sigma-Aldrich

Aluminum sulfate hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
342.15 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
beads or granules
powder, crystals or chunks

grade

for analytical purposes

Assay

98%

form

beads or granules
powder, crystals or chunks

composition

Degree of hydration, 14-18

density

1.69 g/mL at 25 °C (lit.)

SMILES string

[Al+3].[Al+3].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S.H2O/c;;3*1-5(2,3)4;/h;;3*(H2,1,2,3,4);1H2/q2*+3;;;;/p-6

InChI key

BUACSMWVFUNQET-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Aluminum sulfate hydrate is a hydrated aluminum salt. Aluminum sulfate is widely used in oil and fat processing, paper manufacturing and as a flocculant for water treatment. The kinetics of its dehydration by thermal decomposition has been investigated. It is one of the components of hematoxylin solution. Spherical particles of aluminum sulfate hydrate has been synthesized by prilling technique.

Application

Aluminum sulfate hydrate was used in the synthesis of new high crystalline zeolite materials.
It may be used in the synthesis of Al-doped ZnO nanorods and spherical alumina powders.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kinetics of the multistage dehydration of aluminum sulfate hydrate.
Chou KS and Soong CS.
Thermochimica Acta, 81, 305-310 (1984)
Gamma-to-Alpha Transformation in Spherical Aluminum Oxide Powders.
Tucker DS and Bleier A.
Journal of the American Ceramic Society. American Ceramic Society, 68(7), C-163 (1985)
Hydrothermal synthesis of Al-doped ZnO nanorod arrays on Si substrate.
Yun S, et al.
Physica B: Condensed Matter, 405(1), 413-419 (2010)
Prilling of aluminum sulfate hydrates.
Ornek D, et al.
Journal of Chemical Technology and Biotechnology, 75(8), 689-694 (2000)
Structural study on the Al distribution in zeolites Nu-6 (1) and Nu-6 (2).
Galve A et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 145(1), 211-216 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service