Skip to Content
Merck
All Photos(1)

Documents

769460

Sigma-Aldrich

Aluminum hydroxide hydrate

powder, <45 μm

Synonym(s):

Aluminum(III) hydroxide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OH)3 · xH2O
CAS Number:
Molecular Weight:
78.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

particle size

<45 μm

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.01%

SMILES string

O.O[Al](O)O

InChI

1S/Al.4H2O/h;4*1H2/q+3;;;;/p-3

InChI key

SMYKVLBUSSNXMV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum hydroxide hydrate, also known as alumina trihydrate (ATH). It is utilized in a wide variety of applications, serving as an adsorbent, emulsifier, ion-exchanger, and catalyst.

Application

Aluminum hydroxide hydrate can be used:
  • To fabricate Zn anodes for rechargeable aqueous batteries. It helps to form hybrid protective layer to form dendrite free electrode.
  • To prepare flame retardant composites. The addition of ATH led to a substantial decrease in heat and smoke release, as well as a reduction in the generation of harmful volatile compounds during the combustion of asphalt.
  • To fabricate coating material for battery separators for Li-ion batteries.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Surface Modification of LiCo1/3Ni1/3Mn1/3O2 for Lithium Battery Applications
Ganguli, B., et al.
Journal of Electronic Materials, 43(4), 1062-1070 (2014)
New flame-retardant composite separators based on metal hydroxides
for lithium-ion batteries
Yeon, D., et al.
Electrochimica Acta, 157, 282-289 (2015)
Synthesis and Characterization of Al(OH)3, Al2O3 Nanoparticles and Polymeric Nanocomposites
Goudarzi, M., et al.
Journal of Cluster Science, 27(1), 25-38 (2016)
Expanding performance limit of lithium-ion batteries simply by mixing Al(OH)3 powder with LiCoO2
Lee, H., et al.
Journal of Power Sources, 176(1), 359-362 (2008)
Yu Ri Woo et al.
International journal of molecular sciences, 21(17) (2020-08-28)
Air pollution reportedly contributes to the development and exacerbation of atopic dermatitis (AD). However, the exact mechanism underlying this remains unclear. To examine the relationship between air pollution and AD, a clinical, histological, and genetic analysis was performed on particulate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service