Skip to Content
Merck
All Photos(1)

Documents

60700

Sigma-Aldrich

Nuclear Fast Red

for microscopy (Bot., Hist.)

Synonym(s):

4-Amino-9,10-dihydro-1,3-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8NNaO7S
CAS Number:
Molecular Weight:
357.27
Colour Index Number:
60760
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bot., Hist.)

form

powder

solubility

methanol: water (1:1): 0.01 g/10 mL, red

εmax

≥275 at 550-560 nm in DMSO
≥300 at 516-526 nm in DMSO

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Nc1c(O)c(c(O)c2C(=O)c3ccccc3C(=O)c12)S([O-])(=O)=O

InChI

1S/C14H9NO7S.Na/c15-9-7-8(12(18)14(13(9)19)23(20,21)22)11(17)6-4-2-1-3-5(6)10(7)16;/h1-4,18-19H,15H2,(H,20,21,22);/q;+1/p-1

InChI key

IFSXZLJQEKGQAF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Nuclear Fast Red (C.l. 60760) is a substituted hydroxy anthraquinone chromophore that can chelate metal ions. It is also known as kernechtrot and 4-Amino-9,10-dihydro-1,3-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt. It is used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin. It is a dry staining dye that makes target structures (by fixing, wherenecessary embedding, staining with the above nuclear fast red solution, counterstaining, mounting) in human-histological specimen materials, for examplehistological sections of e. g. the kidney, evaluable for diagnostic purposes.

Application

Nuclear fast red has been used for the nuclear staining of PB (Prussian blue) positive cells in the rostral sub-epidermis and upper beak coronal region isolated from pigeons.Recent applications include:
  • staining sperm and spermatids in semen
  • identifying areas for PCR analysis in paraffin-embedded tissues
  • nuclear counterstaining following immunostaining and silver staining
Used as a biological stain

Biochem/physiol Actions

Nuclear fast red dye has anthraquinonic nature and is used either alone or in combination with other dyes. It is mainly used for staining of nuclei.

Principle

Nuclear fast red can be used for simple nuclear staining. It is an acidic dyebelonging to the group of anthraquinone dyes. Aluminum sulfate, functioning as a mordant, is added to form a color lake that enables red nuclear staining. Nuclear fast red-aluminum sulfate is used for counterstaining in histochemicalreactions, e. g. in Berlin blue staining for detection of iron or in silver staining. Nuclear fast red for microscopy Certistain® can also be used fornuclear staining with alcian blue.

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clusters of iron-rich cells in the upper beak of pigeons are macrophages not magnetosensitive neurons.
Treiber CD, et al.
Nature, 484, 367-367 (2012)
Christoph Daniel Treiber et al.
Communicative & integrative biology, 6(4), e24859-e24859 (2013-08-14)
The cells that are responsible for detecting magnetic fields in animals remain undiscovered. Previous studies have proposed that pigeons employ a magnetic sense system that consists of six bilateral patches of magnetite containing dendrites located in the rostral subepidermis of
Kyuri Kim et al.
Journal of ginseng research, 45(1), 98-107 (2021-01-14)
Ginseng extracts and ginseng-fermented products are widely used as functional cosmetic ingredients for their whitening and antiwrinkle effects. Recently, increasing attention has been given to bioactive metabolites isolated from endophytic fungi. However, little is known about the bioactive metabolites of
Kyuri Kim et al.
International journal of molecular sciences, 22(1) (2020-12-31)
With the rapid growth of wireless communication devices, the influences of electromagnetic fields (EMF) on human health are gathering increasing attention. Since the skin is the largest organ of the body and is located at the outermost layer, it is
Exbrayat JM
Genome Visualization by Classic Methods in Light Microscopy, Methods in Visualization (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service