Skip to Content
Merck
All Photos(2)

Key Documents

199656

Sigma-Aldrich

Sudan II

Dye content 90 %

Synonym(s):

1-(2,4-Xylylazo)-2-naphthol, Solvent Orange 7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O
CAS Number:
Molecular Weight:
276.33
Colour Index Number:
12140
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 90%

mp

156-158 °C (lit.)

solubility

chloroform: 10 mg/mL

λmax

493 nm
604 nm (2nd)

ε (extinction coefficient)

≥14000 at 491-497 nm in methanol at 0.005 g/L
≥9000 at 419-425 nm in methanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(\N=N\c2c(O)ccc3ccccc23)c(C)c1

InChI

1S/C18H16N2O/c1-12-7-9-16(13(2)11-12)19-20-18-15-6-4-3-5-14(15)8-10-17(18)21/h3-11,21H,1-2H3/b20-19+

InChI key

JBTHDAVBDKKSRW-FMQUCBEESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sudan II has been used to determine its presence in commercial spices such as hot chilli food samples (LC–ESI–MS/MS).

Biochem/physiol Actions

Sudan II is a fat-soluble synthetic colorant and it belongs to the group of azo compounds. It is classified by the international agency for research on cancer (IARC) as category 3 carcinogens because it has an ability to cause certain forms of liver and bladder cancer in animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

448.9 °F - closed cup

Flash Point(C)

231.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of water-soluble and fat-soluble synthetic colorants in foodstuff by high-performance liquid chromatography-diode array detection-electrospray mass spectrometry.
Ma M
Journal of Chromatography A, 1103, 170-176 (2006)
Determining the adulteration of spices with Sudan I-II-II-IV dyes by UV-visible spectroscopy and multivariate classification techniques.
Di Anibal CV
Talanta, 79, 887-892 (2009)
Shuo Sun et al.
Journal of separation science, 34(14), 1730-1737 (2011-06-10)
The liquid-liquid microextraction (LLME) was developed for extracting sudan dyes from red wine and fruit juice. Room temperature ionic liquid was used as the extraction solvent. The target analytes were determined by high-performance liquid chromatography. The extraction parameters were optimized.
Ting Xu et al.
Analytical biochemistry, 405(1), 41-49 (2010-06-05)
To obtain antibodies to develop an enzyme-linked immunosorbent assay (ELISA) for the analysis of Sudan red I, haptens were designed and synthesized via four different strategies: (i) attachment of a spacer at the para position of the benzene ring, (ii)
Jing Liu et al.
Analytical biochemistry, 423(2), 246-252 (2012-02-14)
Many methods have been reported to determine the residues of Sudan dyes in food samples. Among the reported methods, enzyme-linked immunosorbent assay (ELISA) was a frequently used practical screen tool. In this study, a novel hapten of Sudan 2 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service