Skip to Content
Merck
All Photos(1)

Documents

17373

Sigma-Aldrich

Sudan red G

reagent for Ph. Eur.

Synonym(s):

1-(2-Methoxyphenylazo)-2-naphthol, Oil Red 113

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O2
CAS Number:
Molecular Weight:
278.31
Colour Index Number:
12150
Beilstein:
1843558
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.25

Agency

Ph. Eur.
USP/NF

Quality Level

form

powder

quality

reagent for Ph. Eur.

εmax

≥575 at 489-499 nm in toluene

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COc1ccccc1\N=N\c2c(O)ccc3ccccc23

InChI

1S/C17H14N2O2/c1-21-16-9-5-4-8-14(16)18-19-17-13-7-3-2-6-12(13)10-11-15(17)20/h2-11,20H,1H3/b19-18+

InChI key

ALLOLPOYFRLCCX-VHEBQXMUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karola Luetzow et al.
Journal of biomechanics, 40 Suppl 1, S80-S88 (2007-05-01)
Cell stimulation by bioactive molecules has become an important tool in tissue engineering. The homogeneous incorporation of such molecules within the bulk of a polymer-based scaffold compared to surface coating is considered advantageous for most applications and minimizes a burst
Contact dermatitis due to henna, solvent red 1 and solvent red 3. A case report.
F Wantke et al.
Contact dermatitis, 27(5), 346-347 (1992-11-01)
Han-Wen Sun et al.
Journal of chromatography. A, 1164(1-2), 120-128 (2007-08-19)
An accurate method based on the use of gel permeation chromatography (GPC)-liquid chromatography-tandem mass spectrometry interfaced with electrospray ionization (GPC-LC-ESI-MS/MS) was devised for the simultaneous determination of Sudan (I-IV), Sudan Orange G, Sudan Red B, Sudan Red G, Sudan Red
Yin-Liang Wu et al.
Journal of chromatographic science, 48(1), 63-67 (2010-01-09)
A simple and sensitive high-performance liquid chromatography (HPLC) analytical method for the simultaneous determination of six Sudan dyes (Sudan Red G, Sudan I, Sudan II, Sudan III, Sudan Red 7B, Sudan IV) in animal tissues and eggs was developed. Samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service