Skip to Content
Merck
All Photos(2)

Key Documents

S4136

Sigma-Aldrich

Sudan III

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

1-[4-(Phenylazo)phenylazo]-2-naphthol, Cerasin Red, Fat Ponceau G, Fat Soluble Sudan, Scarlet B, Solvent Red 23, Sudan G, Sudan Red BK, Tony Red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N4O
CAS Number:
Molecular Weight:
352.39
Colour Index Number:
26100
Beilstein:
931185
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Sudan III, certified by the Biological Stain Commission, BioXtra

grade

certified by the Biological Stain Commission

product line

BioXtra

form

powder

composition

Dye content, ≥80%

mp

199 °C (dec.) (lit.)

solubility

toluene: 1 mg/mL, clear, red

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2ccccc2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4

InChI

1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H/b24-23+,26-25+

InChI key

FHNINJWBTRXEBC-QSZPNPOGSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified as a fat staining using the Churukian variant of the Lillie-Ashburn supersaturated isopropanol method. Sudan III is substituted for the oil red O in the procedure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

History of Natto and Its Relatives (1405-2012) null
Bubble size on detachment from the luminal aspect of ovine large blood vessels after decompression: The effect of mechanical disturbance
Respiratory Physiology & Neurobiology (2015)
A Good Manufacturing Practice?grade standard protocol for exclusively human mesenchymal stromal cell?derived extracellular vesicles
karin
Cytotherapy (2017)
Purification of Laboratory Chemicals null
Nicolas Anton et al.
Langmuir : the ACS journal of surfaces and colloids, 25(19), 11413-11419 (2009-10-01)
This paper presents the original method of producing aqueous-core lipid nanocapsule, able to encapsulate both hydrophilic and lipophilic species with relevant yields. The scope offered by the integration of both hydrophilic and lipophilic molecules within the same colloidal objects make

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service