Skip to Content
Merck
All Photos(2)

Key Documents

51383

Supelco

Sudan I

analytical standard

Synonym(s):

1-Phenylazo-2-naphthol, Solvent Yellow 14

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC10H6OH
CAS Number:
Molecular Weight:
248.28
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

131-133 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccc2ccccc2c1N=Nc3ccccc3

InChI

1S/C16H12N2O/c19-15-11-10-12-6-4-5-9-14(12)16(15)18-17-13-7-2-1-3-8-13/h1-11,19H

InChI key

MRQIXHXHHPWVIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>554.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 290 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuande Zhao et al.
Journal of chromatography. A, 1217(45), 6995-7002 (2010-10-01)
A novel molecularly imprinted polymer was synthesized with attapulgite employed as matrix, which is simple and time-saving. In this method, sudan I was chosen as template molecule, 2-vinylpyridine as functional monomer and ethylene glycol dimethacrylate as cross-linking agent, respectively. The
Xinying Ma et al.
Food chemistry, 138(2-3), 739-744 (2013-02-16)
This paper describes a novel electrochemical method for the determination of Sudan I in food samples based on the electrochemical catalytic activity of graphene modified glassy carbon electrode (GMGCE) and the enhancement effect of an anionic surfactant: sodium dodecyl sulphonate
Xiaofeng Yang et al.
Journal of AOAC International, 93(5), 1537-1541 (2010-12-15)
Rapid determination of Sudan I in foodstuffs is very important because it was found to be a carcinogen and its use in the food industry was banned. A rapid, sensitive, and convenient electrochemical method was developed for the determination of
Václav Martínek et al.
Toxicological sciences : an official journal of the Society of Toxicology, 117(2), 359-374 (2010-07-10)
Free radicals generated during peroxidase-catalyzed oxidation of two xenobiotics, carcinogenic Sudan I and an anticancer agent ellipticine, easily attack unmodified proteins but not glycoproteins. A significant inverse correlation between the extent of glycosylation of proteins and the degree of binding
Carolina V Di Anibal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 87, 135-141 (2011-12-14)
Raman spectroscopy combined with multivariate analysis was evaluated as a tool for detecting Sudan I dye in culinary spices. Three Raman modalities were studied: normal Raman, FT-Raman and SERS. The results show that SERS is the most appropriate modality capable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service