Skip to Content
Merck
All Photos(4)

Key Documents

189456

Sigma-Aldrich

Poly(ethylene oxide)

average MV 900,000 (nominal), powder, hydroxyl, BHT as inhibitor

Synonym(s):

Polyethylene oxide, PEO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-CH2CH2O-)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene oxide), average Mv ~900,000 (nominal), powder

form

powder

Quality Level

mol wt

average Mv ~900,000 (nominal)

contains

200-500 ppm BHT as inhibitor

viscosity

8,800-17,600 cP, 5 % in H2O(25 °C, Brookfield)(lit.)

transition temp

Tm 65 °C

density

1.21 g/mL at 25 °C

functional group

hydroxyl

SMILES string

[H]OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene oxide) is a synthetic hydrophilicpolymer available in a number of molecular weights. It can be obtained by thering-opening polymerization of ethylene oxide. This biocompatible polymer can be widely used in the field of biomedical research,tissue engineering and surface functionalization.

Application

PEO can be used to prepare polymer electrolyte systems for energy storage and conversion devices due its low glass transition temperature and ability to form complex with many alkaline salts.

Owing to its hydration and swelling properties, it is widely used as a precursor to prepare block copolymer micelles for selective drug and gene delivery.

Itcan also be used to fabricate nanofibrous membranes for the adsorption of heavymetal ions.

Features and Benefits

  • Excellent polymer chain flexibility
  • Low toxicity
  • High solvation power
  • Lowglass transition temperature

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel O'Brien et al.
Biomedical microdevices, 21(3), 53-53 (2019-06-17)
The biocompatible and biodegradable polymer poly(glycerol sebacate), or PGS, is a rubber-like material that finds use in several biomedical applications. PGS is often cast into a mold to form desired structures; alternatively, blending PGS with other reinforcing polymers produces viscous
Shih-Heng Chen et al.
International journal of molecular sciences, 20(7) (2019-04-04)
Peritendinous adhesion is a major complication after tendon injury and the subsequent repairs or reconstructions. The degree of adhesion can be reduced by the interposition of a membranous barrier between the traumatized tendon and the surrounding tissue. In the present
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
P I Polimeni et al.
Journal of cardiovascular pharmacology, 14(3), 374-380 (1989-09-01)
The acute hemodynamic effects of an intravenously (i.v.) injected poly(ethylene oxide), Polyox WSR N-60K (dose 50 mg/kg), were studied in the open-chest rat anesthetized with sodium pentobarbital. The injectate is one of four drag-reducing polymers known to augment in vitro
M Patel Geeta et al.
Current drug delivery, 6(2), 159-165 (2009-05-20)
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service