Skip to Content
Merck
All Photos(4)

Key Documents

182028

Sigma-Aldrich

Poly(ethylene oxide)

average MV 600,000 (nominal), powder, hydroxyl, BHT as inhibitor

Synonym(s):

Polyethylene oxide, PEO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-CH2CH2O-)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene oxide), average Mv 600,000 (nominal), powder

form

powder

Quality Level

mol wt

average Mv 600,000 (nominal)

contains

200-500 ppm BHT as inhibitor

viscosity

4,500-8,800 cP, 5 % in H2O(25 °C, Brookfield)(lit.)

transition temp

Tm 65 °C

Ω-end

hydroxyl

α-end

hydroxyl

application(s)

battery manufacturing

SMILES string

[H]OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene oxide)(PEO) is a high molecular weight, non-ionic water-soluble polymer. It forms agel on hydration and shows good swelling capacity. PEO polymers are non-toxicand widely used in drug delivery systems to enhance drug solubility.

Application

Poly(ethylene oxide) can be used to prepare:
  • Bioabsorbable and injectable hydrogels for sustained drug release.
  • PEO/graphene oxide composite electrolyte membrane for fuel cells.
  • Poly(ethylene oxide)-b-poly(ε-caprolactone) (PEO-b-PCL) diblock copolymer. Losartan potassium encapsulated (PEO-b-PCL) copolymer can be used as a drug carrier.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angeliki Chroni et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-24)
We report on the preparation of drug nanocarriers by encapsulating losartan potassium (LSR) into amphiphilic block copolymer micelles, utilizing the biocompatible/biodegradable poly(ethylene oxide)-b-poly(ε-caprolactone) (PEO-b-PCL) diblock copolymer. The PEO-b-PCL micelles and LSR-loaded PEO-b-PCL nanocarriers were prepared by organic solvent evaporation method
Jun Li et al.
Journal of biomedical materials research. Part A, 65(2), 196-202 (2003-05-08)
Polymeric hydrogels long have attracted interest for biomaterials applications because of their generally favorable biocompatibility. High in water content, they are particularly attractive for delivery of delicate bioactive agents, such as proteins. However, because they require covalent crosslinking for gelation
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
M Patel Geeta et al.
Current drug delivery, 6(2), 159-165 (2009-05-20)
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a
D D Smyth et al.
Cardiovascular drugs and therapy, 4(1), 297-300 (1990-02-01)
Previous studies have demonstrated that Separan AP-30, a drag-reducing polymer, significantly decreased the formation of atherosclerotic plaques in rabbits fed a high-cholesterol diet. Furthermore, Separan AP-273, a polymer similar to but longer than Separan AP-30, markedly increased cardiac output in

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

See All

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service