Skip to Content
Merck
All Photos(2)

Key Documents

179728

Sigma-Aldrich

Super-Hydride® solution

1.0 M lithium triethylborohydride in THF

Synonym(s):

Lithium triethylborohydride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li(C2H5)3BH
CAS Number:
Molecular Weight:
105.94
Beilstein:
4151240
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M lithium triethylborohydride in THF

density

0.892 g/mL at 25 °C

SMILES string

[Li+].[H][B-](CC)(CC)CC

InChI

1S/C6H16B.Li/c1-4-7(5-2)6-3;/h7H,4-6H2,1-3H3;/q-1;+1

InChI key

WCJAYABJWDIZAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Super-Hydride® solution (Lithium triethylborohydride or LiTEBH) is widely used as a powerful and selective reducing agent that shows super hydride activity in organic synthesis.

Application

LiTEBH can be used as a reagent:
  • To reduce alkyl halides to alkanes via dehydrogenation reactions.
  • For the selective reduction of epoxides to Markovnikov alcohols.
  • To reduce tosylates or mesylates primary alcohols to hydrocarbons.
  • For reductive cyclization reactions for the preparation of useful intermediates.
  • In the synthesis of hepta(manno-3-deoxy-6-O-t-butyldimethylsilyl)-β-cyclodextrin by reduction of hepta(manno-2,3-anhydro-6-O-t-butyldimethylsilyl)-β-cyclodextrin.
  • For hydrodefluorination of C-F bonds using Ni catalyst.
  • To prepare alkynyl alcohols from cleavage of cyclic keto-vinyl triflates.
  • In the stereoselective reduction of bicyclic imides, isoquinolines, and pyridines.
  • To prepare tungsten and molybdenum hydride complexes.

Legal Information

Super-Hydride is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Seven-coordinate hydride complexes of molybdenum and tungsten. Crystal and molecular structures of WH(Cl)(CO)2(PMe3)3
Contreras L, et al.
Organometallics, 12(10), 4228-4228 (1993)
Lithium Triethylborohydride
Zaidlewicz M, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-10 (2001)
Shin Kamijo et al.
Journal of the American Chemical Society, 128(19), 6499-6507 (2006-05-11)
A thorough analysis of the chemistry of vinylogous acyl triflates provides insight into important chemical processes and opens new directions in synthetic technology. Tandem nucleophilic addition/C-C bond cleaving fragmentation reactions of cyclic vinylogous acyl triflates 1 yield a variety of
Sci. Synth., 2, 13-89 (2003)
Regio-and stereoselective reduction of bicyclic imides for the asymmetric synthesis of highly substituted pyrrolidines
Deprez P, et al.
Tetrahedron, 49(18), 3781-3792 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service