Skip to Content
Merck
All Photos(2)

Key Documents

658235

Sigma-Aldrich

Lithium dimethylaminoborohydride solution

1 M in THF

Synonym(s):

N-Methylmethanamine boron complex, Lithium (dimethylamino)trihydroborate, Lithium trihydro(N-methylmethanaminato)borate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H9BLiN
CAS Number:
Molecular Weight:
64.85
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: reductant

Quality Level

concentration

1 M in THF

refractive index

n20/D 1.423

density

0.882 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].[BH3-]N(C)C

InChI

1S/C2H9BN.Li/c1-4(2)3;/h1-3H3;/q-1;+1

InChI key

CEDUMRZWZLVFKS-UHFFFAOYSA-N

General description

Lithium dimethylaminoborohydride is a reagent exhibiting dual properties like a metal hydride and nitrogen nucleophile. It is generally considered as an alternative to lithium aluminum hydride (LAH) for the reduction of carbonyl compounds, amides, and lactams.

Application

Lithium Aminoborohydride (LAB) Reagents

Reactant for:
  • B-H oxidative addition reactions
  • Reduction and amination reactions
  • Reduction of N-alkyl lactams
  • Synthesis of tertiary amine-boranes
Capable of reducing a variety of fuctional groups.
LABs can transfer the amine moiety, as in the case of the reaction with halopyridines and primary alkyl methanesulfonates.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ab initio analysis of lithium dimethylaminoborohydride
Mogali S, et al.
The Journal of Organic Chemistry, 66(7), 2368-2373 (2001)
Lithium aminoborohydrides: powerful, selective, air-stable reducing agents
Pasumansky L, et al.
Organic Process Research & Development, 10(5), 959-970 (2006)
Saikia, P.P.
Synlett, 995-995 (2007)
Pasumansky, L. et al
Aldrichimica Acta, 38, 61-61 (2005)
Fisher, G. B. et al
The Journal of Organic Chemistry, 59, 6378-6378 (1994)

Articles

Lithium aminoborohydride (LAB) reagents are a new class of powerful and selective reagents developed in the laboratory of Professor Bakthan Singaram at the University of California, Santa Cruz.

Related Content

New asymmetric methodology meets the continuous demand for optically active compounds in various fields.

New asymmetric methodology meets the continuous demand for optically active compounds in various fields.

New asymmetric methodology meets the continuous demand for optically active compounds in various fields.

New asymmetric methodology meets the continuous demand for optically active compounds in various fields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service