Skip to Content
Merck
All Photos(2)

Key Documents

149381

Sigma-Aldrich

2,6-Diethylaniline

98%

Synonym(s):

2-Amino-1,3-diethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2C6H3NH2
CAS Number:
Molecular Weight:
149.23
Beilstein:
1423626
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.02 mmHg ( 20 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.545 (lit.)

bp

243 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

SMILES string

CCc1cccc(CC)c1N

InChI

1S/C10H15N/c1-3-8-6-5-7-9(4-2)10(8)11/h5-7H,3-4,11H2,1-2H3

InChI key

FOYHNROGBXVLLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The 2,6-diethylaniline complexes with iodine, as a sigma-acceptor, were studied spectrophotometrically in chloroform, dichloromethane and carbontetrachloride solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urinary biomonitoring for alachlor exposure in commercial pesticide applicators by immunoassay.
R E Biagini et al.
Bulletin of environmental contamination and toxicology, 54(2), 245-250 (1995-02-01)
S J Logusch et al.
Journal of agricultural and food chemistry, 47(5), 2125-2129 (1999-12-20)
Rat liver tissue homogenates were utilized for in vitro enzymatic conversion of 2,6-diethylaniline (DEA) to the important alachlor metabolite 4-amino-3,5-diethylphenyl sulfate (ADEPS), which was also generated as a radiolabeled standard for use in metabolism studies. ADEPS formation in rodents is
A D Kligerman et al.
Mutation research, 441(1), 95-101 (1999-05-04)
Alachlor is a widely used herbicide for which there is significant human exposure, principally through groundwater contamination and inhalation. Because alachlor is purported to be carcinogenic and mutagenic, we initiated studies to determine if induced cytogenetic damage could be used
M Taningher et al.
Toxicology, 85(2-3), 117-122 (1993-12-31)
Alachlor was studied in vivo for its capability to induce DNA damage, as evaluated by the alkaline elution test. The experiments were performed in mouse and rat liver after acute or subacute intraperitoneal or per os administrations of the chemical
R Galati et al.
Anticancer research, 18(2A), 979-982 (1998-06-06)
In order to estimate the environmental risk of the use of Alachlor, experiments on laboratory animals were conducted. Alachlor and 2,6 diethylaniline content in blood serum was quantified. Three groups of male ACI/T rats and C3H/FEJ mice were treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service