Skip to Content
Merck
All Photos(1)

Documents

136190

Sigma-Aldrich

1-Iodo-2-nitrobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NO2
CAS Number:
Molecular Weight:
249.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

288-289 °C (lit.)

mp

49-51 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1I

InChI

1S/C6H4INO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H

InChI key

JXMZUNPWVXQADG-UHFFFAOYSA-N

Application

1-Iodo-2-nitrobenzene was used in one step synthesis of 2-(2-Pyridyl)-3H-indol-3-one N-Oxide. 1-Iodo-2-nitrobenzene was used in the synthesis of 1-(2-Nitrophenyl)-1H-indole in the presence of PEG3400 (poly(ethylene glycol))–Cs2CO3–copper pre-catalyst under microwave activation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dardan Hetemi et al.
Acta chimica Slovenica, dec(4), 818-824 (2018-01-11)
An efficient, versatile and non-destructive in situ method in reaction monitoring using vibrational spectroscopy is described. A Suzuki cross-coupling reaction was monitored in which the substrate 1-iodo-2-nitrobenzene reacted with the electrophile phenylboronic acid to form the product 2-nitrobiphenyl. To hasten
A one-step synthesis of 2-(2-Pyridyl)-3H-indol-3-one N-oxide: is it an efficient spin trap for hydroxyl radical?
G M Rosen et al.
The Journal of organic chemistry, 65(14), 4460-4463 (2000-07-13)
PEG3400-Cs2CO3: an efficient and recyclable microwave-enhanced catalytic system for ligand-free Ullmann arylation of indole and benzimidazole.
Colacino E, et al.
Tetrahedron, 66(21), 3730-3735 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service