Skip to Content
Merck
All Photos(1)

Documents

365424

Sigma-Aldrich

1-Bromo-2-nitrobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NO2
CAS Number:
Molecular Weight:
202.01
Beilstein:
2044109
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

261 °C (lit.)

mp

40-42 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccccc1Br

InChI

1S/C6H4BrNO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H

InChI key

ORPVVAKYSXQCJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Bromo-2-nitrobenzene undergoes palladium[0]-mediated Ullmann cross-coupling reaction with a range of β-halo-enals, -enones or -esters to afford the corresponding β-aryl derivatives. Palladium[0]-mediated Ullmann cross-coupling reaction of 1-bromo-2-nitrobenzene with β-bromo-α,β-unsaturated aldehydes is reported.

Application

1-Bromo-2-nitrobenzene may be used in the preparation of:
  • 4-methoxy-2′-nitrodiphenyl ether
  • 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene
  • 5-hydroxy-3-methoxy-2′-nitrodiphenyl ether

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

4-Hydroxy-2'-Nitrodiphenyl Ether Analogues as Novel Tyrosinase Inhibitors.
Sapkota K, et al.
Bull. Korean Chem. Soc., 31(5), 1319-1319 (2010)
Martin G Banwell et al.
Organic letters, 6(16), 2741-2744 (2004-07-30)
Palladium[0]-mediated Ullmann cross-coupling of 1-bromo-2-nitrobenzene (1 R = H) and its derivatives with a range of beta-halo-enals, -enones, or -esters readily affords the corresponding beta-aryl derivatives, which are converted into the corresponding quinolines, 2-quinolones, phenanthridines, or 6(5H)-phenanthridinones on reaction with
New protocols for the synthesis of 3, 4-annulated and 4-substituted quinolines from ?-bromo-a, ?-unsaturated aldehydes and 1-bromo-2-nitrobenzene or 2-bromoacetanilide.
Some S, et al.
Tetrahedron Letters, 48(20), 3609-3612 (2007)
Luka A Wright et al.
Dalton transactions (Cambridge, England : 2003), 44(16), 7230-7241 (2015-03-20)
The 2-(2′-aniline)-6-imine-pyridines, 2-(C6H4-2′-NH2)-6-(CMe=NAr)C5H3N (Ar = 4-i-PrC6H4 (HL1a), 2,6-i-Pr2C6H3 (HL1b)), have been synthesised via sequential Stille cross-coupling, deprotection and condensation steps from 6-tributylstannyl-2-(2-methyl-1,3-dioxolan-2-yl)pyridine and 2-bromonitrobenzene. The palladium(II) acetate N,N,N-pincer complexes, [{2-(C6H4-2′-NH)-6-(CMe=NAr)C5H3N}Pd(OAc)] (Ar = 4-i-PrC6H4 (1a), 2,6-i-Pr2C6H3 (1b)), can be prepared by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service