Skip to Content
Merck
All Photos(1)

Documents

15200

Sigma-Aldrich

Bis[3-(triethoxysilyl)propyl] tetrasulfide

technical, ≥90% (NMR)

Synonym(s):

TESPTS, 3,3′-Tetrathiobis(propyl-triethoxysilane)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H42O6S4Si2
CAS Number:
Molecular Weight:
538.95
Beilstein:
2062235
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (NMR)

form

liquid

refractive index

n20/D 1.49

density

1.08 g/mL at 20 °C (lit.)

SMILES string

CCO[Si](CCCSSSSCCC[Si](OCC)(OCC)OCC)(OCC)OCC

InChI

1S/C18H42O6S4Si2/c1-7-19-29(20-8-2,21-9-3)17-13-15-25-27-28-26-16-14-18-30(22-10-4,23-11-5)24-12-6/h7-18H2,1-6H3

InChI key

VTHOKNTVYKTUPI-UHFFFAOYSA-N

Application

Bis[3-(triethoxysilyl)propyl] tetrasulfide (TESPTS) is generally used as a crosslinking agent and reinforcing filler in the manufacturing of rubber. TESPT film provides corrosion protection to the metallic substrates. It is also used as a precursor in the synthesis of mesoporous organosilicas.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Modification of bis-silane solutions with rare-earth cations for improved corrosion protection of galvanized steel substrates.
Montemor, MF et al.
Progress in Organic Coatings, 57(1), 67-77 (2006)
Bo Chen et al.
Scientific reports, 5, 9017-9017 (2015-03-13)
There is no effective strategy for the treatment of spinal cord injury (SCI). An appropriate combination of hydrogel materials and neurotrophic factor therapy is currently thought to be a promising approach. In this study, we performed experiments to evaluate the
Elkid Cobani et al.
Nanomaterials (Basel, Switzerland), 9(4) (2019-04-03)
Sepiolite (Sep)⁻styrene butadiene rubber (SBR) nanocomposites were prepared by using nano-sized sepiolite (NS-SepS9) fibers, obtained by applying a controlled surface acid treatment, also in the presence of a silane coupling agent (NS-SilSepS9). Sep/SBR nanocomposites were used as a model to
Thioether-bridged Mesoporous Organosilicas: Mesophase Transformations Induced by the Bridged Organosilane Precursor.
Liu, et al.
Advances in Functional Materials, 17(4), 569-576 (2007)
Kumarjyoti Roy et al.
Polymers, 12(2) (2020-02-13)
In the present study, the suitability of various chemical treatments to improve the performance of jute fibers (JFs) filled natural rubber (NR) composites was explored. The surface of JFs was modified by three different surface treatments, namely, alkali treatment, combined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service