Skip to Content
Merck
All Photos(1)

Documents

452246

Sigma-Aldrich

1,6-Bis(trichlorosilyl)hexane

97%

Synonym(s):

1,6-Bis(trichlorosilyl)hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3Si(CH2)6SiCl3
CAS Number:
Molecular Weight:
353.05
Beilstein:
1768655
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.475 (lit.)

bp

281 °C (lit.)

density

1.327 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)CCCCCC[Si](Cl)(Cl)Cl

InChI

1S/C6H12Cl6Si2/c7-13(8,9)5-3-1-2-4-6-14(10,11)12/h1-6H2

InChI key

ICJGKYTXBRDUMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,6-Bis(trichlorosilyl)hexane is a silane based cross-linking agent that can be used in the synthesis of polymeric blended films. These films are majorly utilized as dielectrics for applications in organic electronic based devices such as organic field effect transistors (OFETs) and organic thin film transistors (OTFTs).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Printable cross-linked polymer blend dielectrics. Design strategies, synthesis, microstructures, and electrical properties, with organic field-effect transistors as testbeds
Kim C, et al.
Journal of the American Chemical Society, 130(21), 6867-6878 (2008)
Ultra-thin polymer gate dielectrics for top-gate polymer field-effect transistors
Noh Y and Sirringhaus H
Organic Electronics, 10(1), 174-180 (2009)
High-performance transparent inorganic-organic hybrid thin-film n-type transistors
Wang L, et al.
Testing, 5(11), 893-893 (2006)
High-capacitance organic nanodielectrics: Effective medium models of their response
DiBenedetto SA, et al.
The Journal of Physical Chemistry B, 110(45), 22394-22399 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service