281778
(3-Aminopropyl)trimethoxysilane
97%
Synonym(s):
3-(Trimethoxysilyl)propylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2N(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
179.29
Beilstein:
2038118
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.424 (lit.)
bp
91-92 °C/15 mmHg (lit.)
density
1.027 g/mL at 25 °C (lit.)
SMILES string
CO[Si](CCCN)(OC)OC
InChI
1S/C6H17NO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-7H2,1-3H3
InChI key
SJECZPVISLOESU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
(3-Aminopropyl) trimethoxysilane (APTMS) is an aminosilane which is used in silanization processes as a silane coupling agent. It functionalizes substrates with alkoxysilane molecules.
(3-Aminopropyl)trimethoxysilane(APTMS) is an amino-silane that is mainly used as a silane coupling agent for the surface modification of a variety of nanomaterials. APTMS based films form a thermally stable layer on different substrates.
Application
APTMS can be multilayered on SiOx substrates by a layer by layer self assembly. APTMS has also been used as a silane coupling agent on silver nanoparticles.
APTMS is used for the synthesis of gold nano-bipyramids which can be used as substrates for C-reactive protein (CRP) antibodies that were detected by localized surface plasmon resonance(LSPR). Mesoporous silica matrix can be incorporated with APTMS for the removal of chromium (Cr) from waste water. It can also be used to silanize magnetic iron oxide nanoparticles(MIONPs) for the separation of cross-linked enzyme aggregations (CLEAs) from the reaction medium. It may be coated on TiO2 to improve the power conversion efficiency (PCE) as a critical parameter to assess the overall performance of heterojunction perovskite solar cells (PSCs).
Packaging
Optically tunable core-shell composite nanorod structures have been prepared by depositing gold onto silica and titania nanorods which have been surface modified with (3-Aminopropyl)trimethoxysilane
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Self-assembly of the 3-aminopropyltrimethoxysilane multilayers on Si and hysteretic current-voltage characteristics
Applied Physics. A, Materials Science & Processing, 90(3), 581-589 (2008)
Investigation of the mechanism of chromium removal in (3-aminopropyl) trimethoxysilane functionalized mesoporous silica.
Lee J, et al
Scientific Reports, 8(12), 103240E-103240E (2018)
Daijun Zhou et al.
International journal of nanomedicine, 13, 4157-4169 (2018-07-27)
This study aimed to prepare an eco-friendly dressing using a chitin-derived membrane with amphipathic anion/quaternary ammonium salt designed for antibacterial purposes. Four dressings were prepared and group A was chitin, group B was chitin + amphiphilic ion, group C was
Reduction of silver nanoparticles in DMF. Formation of monolayers and stable colloids.
Pastoriza S, et al.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 72(1/2), 83-90 (2000)
Yousef Nazari et al.
Environmental science and pollution research international, 26(24), 25359-25371 (2019-07-01)
In the present article, Fe3O4@TiO2 core/shell (FT) linked to graphene was fabricated by sol-gel technique as a photocatalyst and was employed for the solar degradation of cationic methylene blue (MB) in aqueous solution. The prepared core/shells were linked to graphene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service