Skip to Content
Merck
All Photos(1)

Key Documents

440167

Sigma-Aldrich

(3-Glycidyloxypropyl)trimethoxysilane

≥98%

Synonym(s):

3-(2,3-Epoxypropoxy)propyltrimethoxysilane, GLYMO, Glycidyl 3-(trimethoxysilyl)propyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20O5Si
CAS Number:
Molecular Weight:
236.34
Beilstein:
4308125
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.429 (lit.)

bp

120 °C/2 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCOCC1CO1)(OC)OC

InChI

1S/C9H20O5Si/c1-10-15(11-2,12-3)6-4-5-13-7-9-8-14-9/h9H,4-8H2,1-3H3

InChI key

BPSIOYPQMFLKFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(3-Glycidyloxypropyl)trimethoxysilane (GPTMS) is a bifunctional organosilane with three methoxy groups on one side and an epoxy ring on the other. The methoxy groups bind well with glass substrates creating a 3D matrix. The epoxy group is reactive with amides, alcohols, thiols and acids. GPTMS is highly reactive in water and can be used as a linking agent between the surface of the silica and the polymeric matrix.

Application

GPTMS is widely used as a silica precursor. GPTMS alone with tetraethylortosilicate (TEOS) can be blended with chitosan for use as a filler for polymeric scaffold for bone tissue engineering. It can also be coated on the surface of aluminium alloy to protect from corrosion. GPTMS can used to functionalize (wrap) multi-walled carbon nanotubes (MWCNTs) which can be used as s a reinforcement in epoxy matrix nanocomposites.
Pre-treatment of carbon steel with 3-(Glycidoxypropyl)trimethoxysilane enhances the dry and wet adhesion while reducing the cathotic disbondment rate of an epoxy coating

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalization of carbon nanotubes with (3-glycidyloxypropyl)-trimethoxysilane: Effect of wrapping on epoxy matrix nanocomposites.
Hoepfner, et al.
Journal of Applied Polymer Science, 133(47), 814-820 (2016)
Effect of (3-glycidyloxypropyl) trimethoxysilane (GOPS) on the electrical properties of PEDOT: PSS films.
kansson H, et al.
Journal of Polymer Science. Part B, Polymer Physics, 55(10), 814-820 (2017)
Harun, M.K.; et al.
Progress in Organic Coatings, 4, 317-317 (2005)
Ahmed A Nada et al.
International journal of biological macromolecules, 133, 538-544 (2019-04-22)
Homogenous allocation of inorganic particles in a polymeric matrix is a challenge. In this work, we discuss electrospinning of chitosan /gelatin using CS-Si (chitosan silicone hybrids) as well as the formation of homogeneously distributed Zn elements inside chitosan nanofiber through
Room temperature ionic liquids for epoxy nanocomposite synthesis: Direct dispersion and cure.
Throckmorton JA, et al.
Composites Science and Technology, 86(47), 38-44 (2013)

Articles

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service