Merck
  • Identification of KRAS and PIK3CA but not BRAF mutations in patients with gastric cancer.
Identification of KRAS and PIK3CA but not BRAF mutations in patients with gastric cancer.
Molecular medicine reports (2015-03-31)
Weipeng Lu, Hong Wei, Mei Li, Hai Wang, Lina Liu, Qiuping Zhang, Lisha Liu, Shen Lu
ABSTRACT
Cetuximab, an immunoglobulin G1 chimeric monoclonal antibody directed against the epidermal growth factor receptor, is currently considered to be the strategy with the most potential for the treatment of gastric cancer due to the low frequency of KRAS mutations in patients with gastric cancer. However, the therapeutic success of cetuximab in colorectal cancer (CRC) has demonstrated that the clinical effect of cetuximab is closely dependent not only on KRAS mutations, but also BRAF and phosphoinositide-3-kinase, catalytic, α polypeptide (PIK3CA) mutations. In the present study, the status of KRAS, BRAF and PIK3CA mutations in gastric cancer were investigated concomitantly in order to aid the selection of patients eligible for treatment with cetuximab. Mutations in KRAS (exon 2), BRAF (exon 15) and PIK3CA (exon 9 and exon 20) were retrospectively evaluated by high resolution melting analysis and DNA direct sequencing in samples from 156 patients with gastric cancer. Mutations in either KRAS or PIK3CA were identified in 13 samples (8.3%), 7 samples with KRAS mutations and 6 samples with PIK3CA mutations. No mutations in the BRAF gene were identified. The frequency of mutations in either KRAS or PIK3CA were significantly higher in patients without lymph node metastasis than those with. Furthermore, KRAS and PIK3CA mutations were mutually exclusive. The present study, therefore, suggested that it may be necessary to evaluate KRAS and PIK3CA mutations concomitantly for the selection of patients eligible for treatment with cetuximab.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
PI3 kinase (p110a/p85a) Active human, recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm