Merck
  • Amplified electrochemical hydrogen peroxide reduction based on hemin/G-quadruplex DNAzyme as electrocatalyst at gold particles modified heated copper disk electrode.
Amplified electrochemical hydrogen peroxide reduction based on hemin/G-quadruplex DNAzyme as electrocatalyst at gold particles modified heated copper disk electrode.
Biosensors & bioelectronics (2015-06-05)
Shao-Hua Wu, You Tang, Liang Chen, Xian-Gui Ma, Song-Mao Tian, Jian-Jun Sun
ABSTRACT
A new gold particles modified heated copper disk electrode (Au-HCuDE) with direct current was fabricated. The hemin/G-quadruplex horseradish peroxidase-mimicking DNAzyme (HRP-DNAzyme) was self-assembled on the heated electrode and resulted in a new biosensor denoted as HRP-DNAzyme/Au-HCuDE. By controlling the temperature of the surface of the electrode, the dramatic temperature effect on the electrocatalytic reduction of H2O2 at HRP-DNAzyme/Au-HCuDE sensing platform was demonstrated. This electrocatalytic activity of HRP-DNAzyme was enhanced with electrode temperature elevated. This method was thus preliminarily used to develop an electrochemical biosensor for highly sensitive detection of H2O2. A detection limit of 1.6×10(-7) M could be obtained (S/N=3) with an electrode temperature of 50 °C, which was more than one magnitude lower than that at electrode temperature of 0 °C. This heated electrochemical biosensor shows many merits such as easy fabrication and simple heating equipment, low cost, high thermal stability, and high sensitivity and good reproducibility.
Sigma-Aldrich
6-Mercapto-1-hexanol, 97%
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Hemin, from bovine, ≥90%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
6-Mercapto-1-hexanol, 99%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)