Skip to Content
Merck
All Photos(4)

Key Documents

T6666

Sigma-Aldrich

Trizma® hydrochloride

BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)

Synonym(s):

TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.0% (titration)

form

powder

impurities

Insoluble matter, passes filter test

ign. residue (900 °C)

≤0.2% (as SO4)

loss

≤0.2% loss on drying, 110°C

pH

3.5-5.0 (0.5 M in H2O)

useful pH range

7.0-9.0

pKa (25 °C)

8.1

mp

150-152 °C

solubility

H2O: 0.5 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.010 at 280 in H2O at 0.5 M
≤0.015 at 260 in H2O at 0.5 M

application(s)

diagnostic assay manufacturing

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Easily compare specifications for Trizma HCl products with the Trizma HCl specification table.
The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le D He et al.
Bio-protocol, 7(5), e2157-e2157 (2017-03-05)
Drechmeria coniospora is a nematophagous fungus and potential biocontrol agent. It belongs to the Ascomycota. It is related to Hirsutella minnesotensis, another nematophagous fungus but, phylogenetically, it is currently closest to the truffle parasite Tolypocladium ophioglossoides. Together with its natural
Antonio F Melo Junior et al.
Life sciences, 257, 118138-118138 (2020-07-28)
Hypertension is a relevant sex and sex hormones-dependent risk factor where the cardiovascular and renal health of the population are concerned. Men experience greater losses of renal function (RF) than women, but the mechanisms remain somewhat unclear. Our goal was
Lorena Martin-Jaular et al.
The EMBO journal, 40(8), e105492-e105492 (2021-03-13)
Cells release diverse types of extracellular vesicles (EVs), which transfer complex signals to surrounding cells. Specific markers to distinguish different EVs (e.g. exosomes, ectosomes, enveloped viruses like HIV) are still lacking. We have developed a proteomic profiling approach for characterizing
Tibor Maliar et al.
Chemistry & biodiversity, 12(11), 1678-1695 (2015-11-17)
The content of biogenic compounds and the biological activities of barley (Hordeum vulgare L.)-grain extracts was evaluated. The sufficiently large and heterogeneous set of barley genotypes (100 accessions) enabled the selection of special genotypes interesting for potential industrial, pharmaceutical, and
Yingning Zheng et al.
Analytica chimica acta, 834, 45-50 (2014-06-15)
Our present work aimed at developing a pseudo triple-enzyme cascade electrocatalytic electrochemical aptasensor for determination of thrombin with the amplification of alcohol dehydrogenase (ADH)-Pt-Pd nanowires bionanocomposite and hemin/G-quadruplex structure that simultaneously acted as NADH oxidase and HRP-mimicking DNAzyme. With the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service