Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Product Name
HEPES buffer solution, 1 M in H2O
form
liquid
Quality Level
concentration
1 M in H2O, 238 g/L HEPES in H2O
impurities
DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected
pH
5.5±0.5
useful pH range
6.8-8.2
density
1.07 g/mL at 20 °C
application(s)
diagnostic assay manufacturing
SMILES string
OCCN1CCN(CCS(=O)(O)=O)CC1
InChI
1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)
InChI key
JKMHFZQWWAIEOD-UHFFFAOYSA-N
General description
HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.
Application
HEPES has been used:
- As a component of SAPI medium used for the culturing of S. Typhymurium inoculum
- To prepare differentiation medium for PC12 cells. It is also a component of culture medium RPMI
- As a component of dissection buffer for culturing rat cortical neurons
- Along with MgCl2, KCl and Spermine in hypotonic buffer in mGRASP method to detect mammalian synapses
- As a component of Danieu′s aquarium water used in agarose gel preparation
Still not finding the right product?
Explore all of our products under HEPES buffer solution
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Novel membrane-based electrochemical sensor for real-time bio-applications.
Alatraktchi FA, et al.
Sensors, 14(11), 22128-22139 (2014)
Phagocytosis Assay of Microglia for Dead Neurons in Primary Rat Brain Cell Cultures.
Zhao X, et al.
Bio-protocol, 6(8) (2016)
Cultivation of Salmonella enterica serovar Typhimurium in a norepinephrine-containing medium alters in vivo tissue prevalence in swine.
Toscano MJ, et al.
Journal of experimental animal science, 43 , 329-338 (2007)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 83264-500ML-F | 04061833005514 |
| 83264-100ML-F | 04061838254412 |