Merck
  • Determination of solubility parameters of ionic liquids and ionic liquid/solvent mixtures from intrinsic viscosity.
Determination of solubility parameters of ionic liquids and ionic liquid/solvent mixtures from intrinsic viscosity.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-08-26)
Piyarat Weerachanchai, Yuewen Wong, Kok Hwa Lim, Timothy Thatt Yang Tan, Jong-Min Lee
ABSTRACT
The total and partial solubility parameters (dispersion, polar and hydrogen-bonding solubility parameters) of ten ionic liquids were determined. Intrinsic viscosity approaches were used that encompassed a one-dimensional method (1D-Method), and two different three-dimensional methods (3D-Method1 and 3D-Method2). The effect of solvent type, the dimethylacetamide (DMA) fraction in the ionic liquid, and dissolution temperature on solubility parameters were also investigated. For all types of effect, both the 1D-Method and 3D-Method2 present the same trend in the total solubility parameter. The partial solubility parameters are influenced by the cation and anion of the ionic liquid. Considering the effect on partial solubility parameters of the solvent type in the ionic liquid, it was observed that in both 3D methods, the dispersion and polar parameters of a 1-ethyl-3-methylimidazolium acetate/solvent (60:40 vol %) mixture tend to increase as the total solubility parameter of the solvent increases.
Product Description
Pyrrolidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylene carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Nitromethane, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Nitromethane, ACS reagent, ≥95%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Propylene carbonate, suitable for HPLC, 99.7%
Sigma-Aldrich
Nitromethane, suitable for HPLC, ≥96%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Ethyl-3-methylimidazolium acetate, 97%