Skip to Content
Merck
All Photos(1)

Key Documents

230731

Sigma-Aldrich

Nitromethane

ReagentPlus®, ≥99.0%

Synonym(s):

Nitrocarbol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3NO2
CAS Number:
Molecular Weight:
61.04
Beilstein:
1698205
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
101.2 °C (lit.)
vapor pressure:
2.7 mmHg
Pricing and availability is not currently available.

grade

reagent

vapor density

2.1 (vs air)

vapor pressure

2.7 mmHg

product line

ReagentPlus®

Assay

≥99.0%

form

liquid

autoignition temp.

784 °F

expl. lim.

7.3 %, 33 °F

dilution

(for general lab use)

refractive index

n20/D 1.382 (lit.)

pH

6.4 (20 °C, 0.01 g/L)

bp

101.2 °C (lit.)

mp

−29 °C (lit.)

density

1.127 g/mL at 25 °C (lit.)

SMILES string

C[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3

InChI key

LYGJENNIWJXYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitromethane (CH3NO2) is the simplest nitro organic compound used for a wide range of applications, including as polar solvents to racing fuel. It serves as a solvent for organic chemistry and as a valuable building block for various useful compounds like nitroalkanes, β-nitroalcohols, nitroalkenes, carbonyl compounds, amines, and heterocycles. In industry, nitromethane can be used to stabilize halogenated hydrocarbons.

Application

Nitromethane can be used:
  • As a reagent in the synthesis of 3-nitroindoles by arylation with N-(2-bromoaryl) imidates using palladium catalyst.
  • As a cyanating reagent for the synthesis of thiocyanates in presence of base (KOAc) and iodine.
  • In the preparation of cobalt cage complexes from polyamines and formaldehyde.
  • As a solvent in the preparation of β-substituted γ-nitroaldehydes by enantioselective cross-coupling of β-arylated or γ,δ-unsaturated aldehydes using oxidizing agent DDQ (2,3-dichloro-5,6-dicyanoquinone) and catalyst diphenylprolinol silyl ether.

Features and Benefits

Nitromethane as high energy monopropellant exhibits
  • Low toxicity.
  • High specific impulse.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The catalytic chemistry of nitromethane over Co-ZSM5 and other catalysts in connection with the methane-NOxSCR reaction.
Cowan AD, et al.
J. Catal., 176(2), 329-343 (1998)
Metal ion encapsulation: cobalt cages derived from polyamines, formaldehyde, and nitromethane.
Geue RJ, et al.
Journal of the American Chemical Society, 106(19), 5478-5488 (1984)
Benedek Vakulya et al.
Organic letters, 7(10), 1967-1969 (2005-05-07)
Cinchona alkaloid-derived chiral bifunctional thiourea organocatalysts were synthesized and applied in the Michael addition between nitromethane and chalcones with high ee and chemical yields.
Wenguo Yang et al.
Chemistry, an Asian journal, 7(4), 771-777 (2012-02-10)
Through the cleavage of the C-C bond, the first catalytic tandem conjugate addition-elimination reaction of Morita-Baylis-Hillman C adducts has been presented. Various S(N)2'-like C-, S-, and P-allylic compounds could be obtained with exclusive E configuration in good to excellent yields.
Eagleson M.
Concise Encyclopedia Chemistry, 696-696 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service