Skip to Content
Merck
All Photos(2)

Key Documents

B85919

Sigma-Aldrich

2-Butanol

ReagentPlus®, ≥99%

Synonym(s):

sec-Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
Beilstein:
773649
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
98 °C (lit.)
vapor pressure:
12.5 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

2.6 (vs air)

vapor pressure

12.5 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

761 °F

expl. lim.

9.8 %

IVD

for in vitro diagnostic use

dilution

(for general lab use)

refractive index

n20/D 1.397 (lit.)

bp

98 °C (lit.)

mp

−115 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

SMILES string

CCC(C)O

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3

InChI key

BTANRVKWQNVYAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Butanol is secondary alcohol mainly used as a solvent in organic synthesis. 2-butanol readily converts into 2-butanone (methyl ethyl ketone, MEK), which is used as a solvent in the industrial sector and many domestic cleaning products. It is an intermediate in devulcanizing rubber and the production of alkyl ester for use as biodiesel fuel.

Application

2-Butanol is used:
  • As a precursor to produce 2-butanone in presence of KMnO4 oxidant and CPC (N-cetylpyridinium chloride) micellar catalyst.
  • In the production of CH3NH3PbI3 perovskite films.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mark B Shiflett et al.
The journal of physical chemistry. B, 110(29), 14436-14443 (2006-07-21)
In this article, we investigate vapor-liquid-liquid equilibria (VLLE) of binary systems using a simple volumetric method. Being different from the usual cloud-point method for the determination of liquid-liquid separation boundaries, the present volumetric method is able to determine the direct
Dagmara Wojtków et al.
The journal of physical chemistry. A, 110(36), 10552-10557 (2006-09-08)
The effect of temperature and concentration on the structure of sec-butyl alcohol and isobutyl alcohol/water binary mixtures in the alcohol-rich region (mole fraction of water X(H2O) < 0.3) has been studied using Fourier transform (FT) near-infrared (NIR) spectroscopy. The experimental
Fengli Yang et al.
Chemical communications (Cambridge, England), 47(15), 4469-4471 (2011-03-04)
A new solid acid, based on tantalum hydroxide, was used to catalyze saccharide dehydration into 5-hydroxymethylfurfural (HMF) with high catalytic activity and excellent stability in a water-2-butanol biphasic system. Furthermore, good results were also obtained from Jerusalem artichoke juice with
Robert Brosnan et al.
Anesthesia and analgesia, 103(1), 86-91 (2006-06-23)
Chirality has been proposed as a means for distinguishing relevant from irrelevant molecular targets of action, but the sensitivity and specificity of this test is unknown for volatile anesthetics. We applied enantiomers of two chiral anesthetic alcohols (2-butanol and 2-pentanol)
Ravi K Pathak et al.
Environmental science & technology, 46(21), 11660-11669 (2012-09-19)
Limonene has a strong tendency to form secondary organic aerosol (SOA) in the atmosphere and in indoor environments. Initial oxidation occurs mainly via ozone or OH radical chemistry. We studied the effect of O(3) concentrations with or without a OH

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service