Skip to Content
Merck
All Photos(1)

Key Documents

M1775

Sigma-Aldrich

Methanol

Absolute - Acetone free

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

1.11 (vs air)

Quality Level

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

form

liquid
solution

autoignition temp.

725 °F

expl. lim.

36 %

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol is the simplest aliphatic alcohol consisting of methyl and alcohol groups. Methanol is often used as a solvent as well as a reagent in research labs and chemical industries.

Application

Methanol can be used:

  • To prepare methanolic uranyl acetate solution that may be utilized as a stain for tissues.
  • In the synthesis of 4-nitro-N-(quinolin-8-yl) benzenesulfonamide by microwave reaction.

Features and Benefits

Methanol has:
  • Low freezing point.
  • Miscible in water.

Other Notes

Suitable for use as a fixative and in preparing various Romanowsky type staining solutions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AN IMPROVED STAINING METHOD FOR ELECTRON MICROSCOPY.
J G STEMPAK et al.
The Journal of cell biology, 22, 697-701 (1964-09-01)
Nomaan M Rezayee et al.
Journal of the American Chemical Society, 137(3), 1028-1031 (2015-01-17)
This Communication describes the hydrogenation of carbon dioxide to methanol via tandem catalysis with dimethylamine and a homogeneous ruthenium complex. Unlike previous examples with homogeneous catalysts, this CO2-to-CH3OH process proceeds under basic reaction conditions. The dimethylamine is proposed to play
Methanol.
R Von Burg
Journal of applied toxicology : JAT, 14(4), 309-313 (1994-07-01)
CO2 hydrogenation to methanol over CuZnGa catalysts prepared using microwave-assisted methods.
Cai W, et al.
Catalysis Today, 242, 193-199 (2015)
CO2 Hydrogenation to Methanol on Supported Au Catalysts under Moderate Reaction Conditions: Support and Particle Size Effects.
Hartadi Y, et al.
ChemSusChem, 8(3), 456-465 (2015)

Questions

  1. where can I find the expiery date of a bach if it is not present on the COA? I need the information for: methanol M1775, SLCP9620

    1 answer
    1. This product does not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service