Skip to Content
Merck
All Photos(1)

Documents

494437

Sigma-Aldrich

Methanol

≥99.93%, BioReagent

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05

product name

Methanol, BioReagent, ≥99.93%

vapor density

1.11 (vs air)

Quality Level

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

description

gradient analysis <= 0.005 254nm

product line

BioReagent

Assay

≥99.93%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

impurities

≤0.0002 meq/g Titr. base
≤0.0003 meq/g Titr. acid
≤0.02% (water)

evapn. residue

<0.0003%

color

APHA: <10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

solubility

benzene: miscible(lit.)
ethanol: miscible(lit.)
water: miscible(lit.)

density

0.791 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 203 nm Amax: 1.0
λ: 210 nm Amax: 0.60
λ: 220 nm Amax: 0.30
λ: 230 nm Amax: 0.15
λ: 240 nm Amax: 0.05
λ: 260-400 nm Amax: 0.01

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. Shape-selective zeolite-assisted transformation of methanol to C2-C10 hydrocarbons has been studied. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions. Low water content, minimal residue and clean UV spectra are the special characteristics of this solvent. It can be utilized for biotechnological processes like protein sequencing, peptide and oligonucleotide synthesis.

Application

Methanol has been used to fix resolved protein bands on sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) gel.

Preparation Note

Product filtered through a 0.2 μm filter

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Combining Low-Pressure CO2 Capture and Hydrogenation To Form Methanol.
Khusnutdinova JR, et al.
ACS Catalysis, 5(4), 2416-2422 (2015)
Nomaan M Rezayee et al.
Journal of the American Chemical Society, 137(3), 1028-1031 (2015-01-17)
This Communication describes the hydrogenation of carbon dioxide to methanol via tandem catalysis with dimethylamine and a homogeneous ruthenium complex. Unlike previous examples with homogeneous catalysts, this CO2-to-CH3OH process proceeds under basic reaction conditions. The dimethylamine is proposed to play
Methanol.
R Von Burg
Journal of applied toxicology : JAT, 14(4), 309-313 (1994-07-01)
Hong Jiang et al.
Current protocols in chemical biology, 4(1), 19-34 (2012-11-29)
Poly(ADP-ribose) polymerases (PARPs) play important roles in various biological processes, including DNA repair, transcriptional regulation, mitosis, and RNA processing. PARP inhibitors are in clinical trials for treating human cancers. Understanding the biological function of PARPs will be important to fully
CO2 Hydrogenation to Methanol on Supported Au Catalysts under Moderate Reaction Conditions: Support and Particle Size Effects.
Hartadi Y, et al.
ChemSusChem, 8(3), 456-465 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service