Merck
  • Identification of manipulated variables for a glycosylation control strategy.
Identification of manipulated variables for a glycosylation control strategy.
Biotechnology and bioengineering (2014-04-15)
Melissa M St Amand, Devesh Radhakrishnan, Anne S Robinson, Babatunde A Ogunnaike
ABSTRACT
N-linked glycan distribution affects important end-use characteristics such as the bioactivity and efficacy of many therapeutic proteins, (including monoclonal antibodies), in vivo. Yet, obtaining desired glycan distributions consistently during batch-to-batch production can be challenging for biopharmaceutical manufacturers. While an appropriately implemented on-line glycosylation control strategy during production can help to ensure a consistent glycan distribution, to date no such strategies have been reported. Our goal is to develop and validate a comprehensive strategy for effective on-line control of glycosylation, the successful achievement of which requires first identifying appropriate manipulated variables that can be used to direct the glycan distribution to a desired state. While various culture conditions such as bioreactor process variables, media type, and media supplements have been shown to affect the glycan distribution, in this study we focus on the latter. Specifically, we implemented a statistically designed series of experiments to determine the significant main effects (as well as interaction effects) of media supplementation with manganese, galactose, ammonia and found that each had significant effects on certain glycans. We also include data indicating the glycosylation enzyme gene transcript levels as well as the intracellular nucleotide sugar concentrations in the presence of the media supplements to provide insight into the intracellular conditions that may be contributing to the changes in glycan distribution. The acquired experimental data sets were then used to identify which glycans can be controlled by the media supplements and to what degree. We determined that MnCl2 can be used as a manipulated variable to increase the relative abundance of M51 and decrease FA2 simultaneously, and galactose can be used as a manipulated variable to increase the relative abundance of FA2G1 and decrease FA2 and A2 simultaneously.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, anhydrous, free-flowing, Redi-Dri, 97%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)