Skip to Content
Merck
All Photos(1)

Key Documents

98479

Supelco

Tetrabutylammonium bisulfate solution

suitable for ion pair chromatography, LiChropur, concentrate, ampule

Synonym(s):

Tetrabutylammonium hydrogen sulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
Beilstein:
3599663
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

form

solution

quality

LiChropur
ampule

packaging

ampule of 25 mL

technique(s)

ion pair chromatography: suitable

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

General description

Concentrate for 6x1 L ready-to-use solution. Dilute contents of one ampule (~0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenfeng He et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10043-10051 (2012-05-29)
A new kind of organic-inorganic hybrid complexes based on polyoxometalate were synthesized through symmetrically grafting two adeninyl groups onto Anderson-type MnMo(6) clusters and encapsulating the clusters by organic surfactants. The resultant complexes exhibited thermal-induced dynamic self-assembly behaviors which greatly depended
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind
Toshihiro Kimura et al.
Inorganic chemistry, 51(23), 13001-13008 (2012-11-15)
A simple monomeric tungstate, TBA(2)[WO(4)] (I, TBA = tetra-n-butylammonium), could act as an efficient homogeneous catalyst for chemical fixation of CO(2) with 2-aminobenzonitriles to quinazoline-2,4(1H,3H)-diones. Various kinds of structurally diverse 2-aminobenzonitriles could be converted into the corresponding quinazoline-2,4(1H,3H)-diones in high
Melissa M St Amand et al.
Biotechnology and bioengineering, 111(10), 1957-1970 (2014-04-15)
N-linked glycan distribution affects important end-use characteristics such as the bioactivity and efficacy of many therapeutic proteins, (including monoclonal antibodies), in vivo. Yet, obtaining desired glycan distributions consistently during batch-to-batch production can be challenging for biopharmaceutical manufacturers. While an appropriately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service