Skip to Content
Merck
All Photos(2)

Key Documents

311421

Sigma-Aldrich

Perchloric acid

70%, 99.999% trace metals basis

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

6.8 mmHg ( 25 °C)

Assay

70%
99.999% trace metals basis

form

liquid

reaction suitability

reagent type: oxidant

concentration

≥70-- < 90%

bp

ca.203 °C/at 1.013 hPa

mp

18 °C

solubility

water: miscible (completely)

density

1.664 g/mL at 25 °C

cation traces

Al: ≤0.3 ppm
Ca: ≤0.3 ppm
Fe: ≤ 0.1 ppm
Na: 3.1 ppm
Zn: ≤ 0.1 ppm

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perchloric acid (HClO4) can be used in the preparation of:
  • Silica supported perchloric acid (HClO4-SiO2) which is used as a catalyst in geminal diacylation of aldehydes, synthesis of amidoalkyl naphthols, primary carbamates, and tetrasubstituted imidazoles.
  • Alumina supported perchloric acid (Al2O3-HClO4), which is used as a catalyst in the synthesis of α-(α-amidobenzyl)-β-naphthols.
  • Fe3O4@SiO2-HClO4 catalyst, which is used in the synthesis of 1,4-dihydropyridine derivatives.

Perchloric acid can be used as:      
  •   A reaction medium in oxidation reactions.       
  • A reagent for the preparation of fluorinated polymer electrolytes using poly (vinylidenefluoride-cohexafluoropropylene) (PVDF-HFP) for fuel cell applications.        
  • A dopant in the preparation of perchloric acid-functionalized polyaniline (HClO4/PANI) catalyst for the synthesis of 2-substituted benzothiazoles by condensation reaction between o-aminothiophenol with aldehydes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silica supported perchloric acid (HClO4-SiO2): an efficient reagent for the preparation of primary carbamates under solvent-free conditions
Modarresi-Alam AR, et al.
Tetrahedron, 63(36), 8723-8726 (2007)
Kinetics of oxidation of pantothenic acid by chloramine-T in perchloric acid and in alkaline medium catalyzed by OsO4: A mechanistic approach
Jagadeesh RV
International Journal of Chemical Kinetics, 37(4), 201-210 (2005)
Preparation and Application of Perchloric Acid Supported on Alumina (Al2O3-HClO4) to the Synthesis of α-(α-Amidobenzyl)-β-naphthols
Shaterian H, et al.
Chin. J. Chem., 27(4), 815-820 (2009)
Silica supported perchloric acid (HClO4-SiO2): A highly efficient and reusable catalyst for geminal diacylation of aldehydes under solvent-free conditions
Khan AT, et al.
J. Mol. Catal. A: Chem., 255(1-2), 230-235 (2006)
Perchloric acid-doped polyaniline as an efficient and reusable catalyst for the synthesis of 2-substituted benzothiazoles
Abdollahi-Alibeik M, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 184(12), 3182-3190 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service