Skip to Content
Merck
All Photos(2)

Key Documents

86853

Supelco

Tetrabutylammonium bisulfate

suitable for ion pair chromatography, LiChropur, ≥99.0%

Synonym(s):

Tetrabutylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
Beilstein:
3599663
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21
Assay:
≥99.0% (T)
≥99.0%
form:
crystals

description

cationic

Quality Level

Assay

≥99.0% (T)
≥99.0%

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

mp

171-174 °C

anion traces

bromide (Br-): ≤50 mg/kg

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.06
λ: 220 nm Amax: 0.05
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutyammonium bisulfate (TBAHS) is an ion-pairing reagent. It acts as a catalyst in hydrolysis of aziridines and epoxides and the catalyst can be recycled.

Application

TBAHS was suitable as ion-pairing reagent in determination and separation of thallium using ionic liquid-assisted ion-pairing liquid-liquid microextraction and inductively coupled plasma mass spectrometry. It was suitable as mobile phase component for a HPLC-UV analysis to quantify chondroitin sulfate.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

348.1 °F - closed cup

Flash Point(C)

175.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leticia B Escudero et al.
Journal of hazardous materials, 244-245, 380-386 (2012-12-29)
A fast and simple method involving separation and determination of thallium (Tl) species, based on novel ionic liquid-assisted ion pairing dispersive liquid-liquid microextraction (DLLME) method, was developed. Initially, Tl(III) was selectively complexed with chloride ion to form [TlCl(4)](-) chlorocomplex. Subsequently
David Ji et al.
Journal of AOAC International, 90(3), 659-669 (2007-06-22)
A method to quantify chondroitin sulfate in raw materials and dietary supplements at a range of about 5 to 100% (w/w) chondroitin sulfate has been developed and validated. The chondroitin sulfate is first selectively hydrolyzed by chondroitinase ACII enzyme to
Ren-Hua Fan et al.
Organic & biomolecular chemistry, 1(9), 1565-1567 (2003-08-21)
Bu4NHSO4 (TBAHS) is an effective catalyst for the hydrolysis of aziridines and epoxides under mild and non-metal conditions to give the corresponding beta-amino alcohols and 1,2-diols in high yields. The catalyst can be recycled.
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service