Skip to Content
Merck
All Photos(3)

Key Documents

S1888

Sigma-Aldrich

Sucrose

Sucrose

≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

sugar cane

Quality Level

product line

BioReagent

Assay

≥99.5% (GC)

form

crystals

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

impurities

≤5 ppm heavy metals (as lead)

color

white

useful pH range

5.5-7 (25 °C, 342 g/L)

mp

185-187 °C (lit.)

solubility

H2O: 500 mg/mL

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sucrose is an abundant disaccharide produced as the end product of photosynthesis. It is a nonreducing sugar comprising glucose and fructose. Sucrose is obtained from cane sugar and is mostly synthesized in leaves. 

Application

Sucrose has been used as a component of vitrification solutions for the freezing, thawing, or heating of embryo/oocytes. It has also been used to create sucrose gradients for the purification of viruses and proteins.

Biochem/physiol Actions

Sucrose is the chief sugar transferred in the phloem of most plants. It is used to make density gradients for the separation of cell/organelle. Sucrose can also be used in various enzymatic assays. Additionally, it serves as a supplement in the plant, insect, and bacterial culture media.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Harrison et al.
Pediatrics, 130(5), 918-925 (2012-10-10)
The use of oral sucrose has been the most extensively studied pain intervention in newborn care to date. More than 150 published studies relating to sweet-taste-induced calming and analgesia in human infants have been identified, of which 100 (65%) include
Rafaela Ribeiro Reis et al.
Plant science : an international journal of experimental plant biology, 221-222, 59-68 (2014-03-25)
Drought is one of the most challenging agricultural issues limiting sustainable sugarcane production and, in some cases, yield losses caused by drought are nearly 50%. DREB proteins play vital regulatory roles in abiotic stress responses in plants. The transcription factor
William J Armour et al.
The Plant cell, 27(9), 2484-2500 (2015-08-25)
Lobe development in the epidermal pavement cells of Arabidopsis thaliana cotyledons and leaves is thought to take place via tip-like growth on the concave side of lobes driven by localized concentrations of actin filaments and associated proteins, with a predicted
Zsuzsanna Tucsek et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 69(10), 1212-1226 (2013-11-26)
There is growing evidence that obesity has deleterious effects on the brain and cognitive function in the elderly population. However, the specific mechanisms through which aging and obesity interact to promote cognitive decline remain unclear. To test the hypothesis that
Julia B Cordero et al.
The EMBO journal, 33(13), 1474-1491 (2014-05-03)
The non-receptor tyrosine kinase c-Src, hereafter referred to as Src, is overexpressed or activated in multiple human malignancies. There has been much speculation about the functional role of Src in colorectal cancer (CRC), with Src amplification and potential activating mutations

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service