Merck
  • Selective extraction of antimycotic drugs from sludge samples using matrix solid-phase dispersion followed by on-line clean-up.
Selective extraction of antimycotic drugs from sludge samples using matrix solid-phase dispersion followed by on-line clean-up.
Analytical and bioanalytical chemistry (2014-09-28)
Jorge Casado, Gabriela Castro, Isaac Rodríguez, María Ramil, Rafael Cela
ABSTRACT
An effective and selective, modular sample preparation method for the extraction of eight antimycotic drugs, belonging to three different chemical classes, from digested sludge samples is proposed. To this end, matrix solid-phase dispersion (MSPD) was on-line connected with a cationic exchanger solid-phase extraction (SPE) cartridge. Analytes were extracted from the MSPD syringe, which contained the freeze-dried sludge sample dispersed with C18 plus a clean-up layer of primary and secondary amine (PSA) sorbent, with 10 mL of methanol. This extract flowed also through the SPE cartridge, where target compounds remained trapped while neutral interferences are released. After discarding the MSPD syringe, analytes were recovered with 10 mL of methanol (0.5% in NH3) before LC-MS/MS determination using a hybrid quadrupole time-of-flight (QTOF) mass spectrometer furnished with an electrospray ionization (ESI) source. In comparison with previously published sample preparation methodologies, the developed approach greatly simplifies sample handling and reduces attenuation of ESI ionization for sample extracts when compared to standard solutions. The obtained absolute recoveries ranged between 70 and 118%, and the limits of quantification (LOQs) of the method varied between 5 and 8 ng g(-1). Four antimycotic drugs were ubiquitous in urban sludge samples, with maximum average concentrations (above 400 ng g(-1)) corresponding to clotrimazole (CTZ). The screening capabilities of the LC-QTOF-MS system demonstrated that the developed modular extraction and purification methodology might be useful for the selective extraction of other basic drugs (e.g., sertraline, amitryptiline, and amiodarone) from sludge.
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, for inorganic trace analysis, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol