Skip to Content
Merck
All Photos(1)

Documents

09691

Sigma-Aldrich

Ammonium acetate solution

BioUltra, for molecular biology, ~5 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

~5 M in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.01% (as SO4)

pH

6.2-7.5 (25 °C, 1 M in H2O)

density

1.07 g/mL at 20 °C

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.015
λ: 280 nm Amax: 0.01

SMILES string

CC([O-])=O.[H][N+]([H])([H])[H]

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium acetate solution can be used to study molecular biology, biological buffers, reagents and DNA and RNA purification. Ammonium acetate solution has been used to study pharmacokinetic analysis of α and β epimers of glycyrrhetinic acid in rat plasma. Ammonium acetate solution has also been used in a study to develop a method for the simultaneous determination of aristolochic acids A and B in some Chinese herbals and traditional Chinese patent medicines.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Annaelle Soubieux et al.
Journal of oncology pharmacy practice : official publication of the International Society of Oncology Pharmacy Practitioners, 26(2), 413-422 (2019-08-03)
The main objective was to determine the efficacy of various types of cleaning equipment and products after deliberate contamination with cyclophosphamide. The secondary objective was to test various cleaning scenarios using these equipment and products. The study had two phases:
Kerstin Berer et al.
Scientific reports, 8(1), 10431-10431 (2018-07-12)
The autoimmune neurological disease, Multiple Sclerosis (MS), have increased at alarming rates in the Western society over the last few decades. While there are numerous efforts to develop novel treatment approaches, there is an unmet need to identify preventive strategies.
Jessica Z Bereszczak et al.
Protein science : a publication of the Protein Society, 23(7), 884-896 (2014-04-10)
Hepatitis B virus core-antigen (capsid protein) and e-antigen (an immune regulator) have almost complete sequence identity, yet the dimeric proteins (termed Cp149d and Cp(-10)149d , respectively) adopt quite distinct quaternary structures. Here we use hydrogen deuterium exchange-mass spectrometry (HDX-MS) to
Xumin Zhang et al.
Analytical chemistry, 84(18), 7785-7792 (2012-08-15)
Untargeted liquid chromatography-mass spectrometry (LC-MS)-based metabolomics studies are usually carried out in both positive and negative ion modes; however, it is frequently ignored that the optimal conditions in positive ion mode and negative ion mode are often not the same.
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service