A2706
Ammonium acetate solution
for molecular biology, 7.5 M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3COONH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
grade
for molecular biology
Quality Level
sterility
0.2 μm filtered
form
solution
concentration
7.5 M
foreign activity
DNase and RNase, none detected
storage temp.
2-8°C
SMILES string
CC([O-])=O.[H][N+]([H])([H])[H]
InChI
1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
InChI key
USFZMSVCRYTOJT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ammonium acetate solution has been used:
- to prepare mobile phases A and B for sphingolipid liquid chromatography-mass spectrometry (LC-MS) analyse
- for RNA polyA tailing, cDNA synthesis, and digestion of RNA
- in the assessment of in vitro mitochondrial copy number in C9orf72 amyotrophic lateral sclerosis motor neurons
Preparation Note
Prepared in 18 megohm water
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Johan Bielecki et al.
Science advances, 5(5), eaav8801-eaav8801 (2019-05-07)
The possibility of imaging single proteins constitutes an exciting challenge for x-ray lasers. Despite encouraging results on large particles, imaging small particles has proven to be difficult for two reasons: not quite high enough pulse intensity from currently available x-ray
Shi Hu et al.
Oncotarget, 6(3), 1695-1706 (2015-01-31)
Human epidermal growth factor receptors (HERs or ErbBs) play crucial roles in numerous cellular processes. ErbB2 is a key member of ErbB family, and its overexpression is recognized as a frequent molecular abnormality. In cancer, this overexpression correlates with aggressive
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central
Lucie Nováková et al.
Analytica chimica acta, 853, 637-646 (2014-12-04)
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes.
Suzanne L Parker et al.
Journal of pharmaceutical and biomedical analysis, 105, 39-45 (2014-12-22)
A high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method, using hydrophilic interaction liquid chromatography (HILIC) chromatography for the analysis of fosfomycin in human plasma and urine, has been developed and validated. The plasma method uses a simple protein precipitation using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service