Skip to Content
Merck
All Photos(1)

Key Documents

09682

Sigma-Aldrich

Ammonia

puriss., anhydrous, ≥99.95%

Synonym(s):

Ammonia gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
Beilstein:
3587154
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

0.6 (vs air)

Quality Level

vapor pressure

8.75 atm ( 21 °C)

grade

puriss.

Assay

≥99.95%

autoignition temp.

1203 °F

expl. lim.

25 %

impurities

≤0.01% water

bp

−33 °C (lit.)

mp

−78 °C (lit.)

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonia gas is a common reagent or reactant in the synthesis of variety of nitrogen containing inorganic compounds and organic intermediates. It is either used directly in gas phase reaction or bubbled through the reaction mixture.[1]
  • Ammonia can be used in the direct synthesis of primary amines from alcohols.[2]
  • Synthesis of N-doped graphene by vapor deposition in the presence of ammonia gas has been reported.[3][4]

Packaging

Cylinder with net ~1 kg

Other Notes

Sales restrictions may apply

Recommended products

Recommended DIN 477 - Nr. 6

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Gas 2 - Press. Gas Liquefied gas - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonia.
Ullmann's Encyclopedia of Industrial Chemistry (2006)
Selective synthesis of primary amines directly from alcohols and ammonia.
Gunanathan C and Milstein D
Angewandte Chemie (International Edition in English), 47(45), 8661-8664 (2008)
Synthesis of N-doped graphene by chemical vapor deposition and its electrical properties.
Wei D, et al.
Nano Letters, 9(5), 1752-1758 (2009)
Liangti Qu et al.
ACS nano, 4(3), 1321-1326 (2010-02-17)
Nitrogen-doped graphene (N-graphene) was synthesized by chemical vapor deposition of methane in the presence of ammonia. The resultant N-graphene was demonstrated to act as a metal-free electrode with a much better electrocatalytic activity, long-term operation stability, and tolerance to crossover
Olivier Braissant et al.
Journal of inherited metabolic disease, 36(4), 595-612 (2012-10-31)
Hyperammonemia can be caused by various acquired or inherited disorders such as urea cycle defects. The brain is much more susceptible to the deleterious effects of ammonium in childhood than in adulthood. Hyperammonemia provokes irreversible damage to the developing central

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service