Merck
  • Bacterial effector modulation of host E3 ligase activity suppresses PAMP-triggered immunity in rice.
Bacterial effector modulation of host E3 ligase activity suppresses PAMP-triggered immunity in rice.
Nature communications (2014-11-13)
Kazuya Ishikawa, Koji Yamaguchi, Kazuaki Sakamoto, Satomi Yoshimura, Kento Inoue, Seiji Tsuge, Chojiro Kojima, Tsutomu Kawasaki
ABSTRACT
Pathogen effector proteins are delivered to host cells to suppress plant immunity. However, the mechanisms by which effector proteins function are largely unknown. Here we show that expression of XopP(Xoo), an effector of rice pathogen Xanthomonas oryzae pv. oryzae, in rice strongly suppresses peptidoglycan (PGN)- and chitin-triggered immunity and resistance to X. oryzae. XopP(Xoo) targets OsPUB44, a rice ubiquitin E3 ligase with a unique U-box domain. We find that XopP(Xoo) directly interacts with the OsPUB44 U-box domain and inhibits ligase activity. Two amino-acid residues specific for the OsPUB44 U-box domain are identified, which are responsible for the interaction with XopP(Xoo). Silencing of OsPUB44 suppresses PGN- and chitin-triggered immunity and X. oryzae resistance, indicating that OsPUB44 positively regulates immune responses. Thus, it is likely that XopP(Xoo) suppresses immune responses by directly interacting with and inhibiting a positive regulator of plant immunity.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, 2 M in H2O
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Pricing and availability is not currently available.
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Pricing and availability is not currently available.