Merck
  • Environmental contaminants activate human and polar bear (Ursus maritimus) pregnane X receptors (PXR, NR1I2) differently.
Environmental contaminants activate human and polar bear (Ursus maritimus) pregnane X receptors (PXR, NR1I2) differently.
Toxicology and applied pharmacology (2015-02-15)
Roger Lille-Langøy, Jared V Goldstone, Marte Rusten, Matthew R Milnes, Rune Male, John J Stegeman, Bruce Blumberg, Anders Goksøyr
ABSTRACT
Many persistent organic pollutants (POPs) accumulate readily in polar bears because of their position as apex predators in Arctic food webs. The pregnane X receptor (PXR, formally NR1I2, here proposed to be named promiscuous xenobiotic receptor) is a xenobiotic sensor that is directly involved in metabolizing pathways of a wide range of environmental contaminants. In the present study, we comparably assess the ability of 51 selected pharmaceuticals, pesticides and emerging contaminants to activate PXRs from polar bears and humans using an in vitro luciferase reporter gene assay. We found that polar bear PXR is activated by a wide range of our test compounds (68%) but has a slightly more narrow ligand specificity than human PXR that was activated by 86% of the 51 test compounds. The majority of the agonists identified (70%) produces a stronger induction of the reporter gene via human PXR than via polar bear PXR, however with some notable and environmentally relevant exceptions. Due to the observed differences in activation of polar bear and human PXRs, exposure of each species to environmental agents is likely to induce biotransformation differently in the two species. Bioinformatics analyses and structural modeling studies suggest that amino acids that are not part of the ligand-binding domain and do not interact with the ligand can modulate receptor activation.
Product Description
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lindane, PESTANAL®, analytical standard
Supelco
Methoxychlor, PESTANAL®, analytical standard
Supelco
Vinclozolin, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachlorophenol, 97%
Sigma-Aldrich
1,2,4-Trichlorobenzene, anhydrous, ≥99%
Supelco
1,2,4-Trichlorobenzene, PESTANAL®, analytical standard
Supelco
Dieldrin, PESTANAL®, analytical standard
Supelco
Pentachlorophenol, analytical standard
Sigma-Aldrich
1,2,3-Trichlorobenzene, 99%
Sigma-Aldrich
Hexamethylcyclotrisiloxane, 98%
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Hexamethylcyclotrisiloxane, analytical standard
Supelco
Lindane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine, analytical standard
Sigma-Aldrich
1,2,4-Trichlorobenzene, suitable for HPLC, ≥99%
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2,4-Trichlorobenzene, ReagentPlus®, ≥99%
Supelco
Omeprazole, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Clotrimazole, European Pharmacopoeia (EP) Reference Standard
USP
Clotrimazole, United States Pharmacopeia (USP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water