Skip to Content
Merck
All Photos(1)

Documents

48555-U

Supelco

Pentachlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Cl5OH
CAS Number:
Molecular Weight:
266.34
Beilstein:
1285380
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

9.2 (vs air)

vapor pressure

40 mmHg ( 211.2 °C)

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

310 °C (lit.)

mp

165-180 °C (lit.)

density

1.978 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

IZUPBVBPLAPZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentachlorophenol (PCP) is a chlorinated hydrocarbon, an important industrial chemical and a general biocide. It is primarily used in wood protection.

Application

Pentachlorophenol may be used as an analytical reference standard for the determination of the analyte in biological and water samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of pentachlorophenol in urine: the importance of hydrolysis.
Edgerton TR and Moseman RF
Journal of Agricultural and Food Chemistry, 27(1), 197-199 (1979)
Determination of pentachlorophenol in water samples by capillary zone electrophoresis.
Turnes MI, et al.
Journal of Chromatography A, 733(1-2), 395-404 (1996)
Satyendra Kumar Garg et al.
Environmental science and pollution research international, 20(4), 2288-2304 (2012-08-07)
It is the first report in which a novel psychrotrophic Pseudomonas putida SKG-1 strain was evaluated for simultaneous bioremediation of pentachlorophenol and Cr(6+) under various cultural and nutritional conditions. Pentachlorophenol (PCP) dechlorination products, bacterial structure, and functional groups were characterized
Min Yang et al.
Journal of hazardous materials, 237-238, 63-70 (2012-09-12)
In this study, an imprinted silica matrix of pentachlorophenol (PCP) co-loaded with Fe(3)O(4) nanoparticles and ZnS:Mn(2+) quantum dots (QDs) was fabricated. The introduction of Fe(3)O(4) nanoparticles to the imprinted matrix provided an easy way to separate PCP under an external
Weiwei Zheng et al.
Environment international, 42, 105-116 (2011-05-24)
Pentachlorophenol (PCP) has been widely used for killing snails in areas of China where schistosomiasis is epidemic. With the re-emergence of schistosomiasis, the warranted production and consumption of PCP has inevitably resulted in persistent environmental contamination by it and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service