Skip to Content
Merck
All Photos(4)

Key Documents

T54402

Sigma-Aldrich

1,2,3-Trichlorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3
CAS Number:
Molecular Weight:
181.45
Beilstein:
956882
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.25 (vs air)

Quality Level

vapor pressure

0.07 mmHg ( 25 °C)

Assay

99%

autoignition temp.

1059 °F

expl. lim.

6.6 %

bp

218-219 °C (lit.)

mp

51-53 °C (lit.)

SMILES string

Clc1cccc(Cl)c1Cl

InChI

1S/C6H3Cl3/c7-4-2-1-3-5(8)6(4)9/h1-3H

InChI key

RELMFMZEBKVZJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anna K Duell et al.
Chemical research in toxicology, 32(6), 1241-1249 (2019-05-14)
Electronic cigarette liquids (e-liquids) with sweetener additives such as sucralose, a synthetic chlorinated disaccharide, are popular among some e-cigarette consumers; sucralose can be added either by the manufacturer or by the consumer. The prevalence of sucralose in commercial e-liquids is
Qiaoli Liang et al.
Journal of the American Society for Mass Spectrometry, 28(3), 409-418 (2016-12-08)
A new matrix-assisted laser desorption ionization (MALDI) mass spectrometry matrix is proposed for molecular mass determination of polymers. This matrix contains an iron oxide nanoparticle (NP) core with citric acid (CA) molecules covalently bound to the surface. With the assistance
J I Cacho et al.
Talanta, 195, 796-799 (2019-01-11)
A device for headspace sorptive extraction (HSSE) combined with gas chromatography separation and mass spectrometry detection (GC-MS) is presented. The gadget is based on a simple magnetic disk that permits the stir bar to be placed at the top of
Arijit Mukherjee et al.
IUCrJ, 1(Pt 4), 228-239 (2014-07-31)
Aniline-phenol recognition is studied in the crystal engineering context in several 1:1 cocrystals that contain a closed cyclic hydrogen-bonded [⋯O-H⋯N-H⋯]2 tetramer supramolecular synthon (II). Twelve cocrystals of 3,4,5- and 2,3,4-trichlorophenol with one of eight halogenated anilines have been characterized. Ten
Roger Lille-Langøy et al.
Toxicology and applied pharmacology, 284(1), 54-64 (2015-02-15)
Many persistent organic pollutants (POPs) accumulate readily in polar bears because of their position as apex predators in Arctic food webs. The pregnane X receptor (PXR, formally NR1I2, here proposed to be named promiscuous xenobiotic receptor) is a xenobiotic sensor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service